Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP2
Entrez Gene ID
6452
Gene Name
SH3 domain binding protein 2
Gene Aliases
3BP-2, 3BP2, CRBM, CRPM, RES4-23
Chromosome Location
Chr.4: 2793023 - 2841096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122681.1 NP_001116153.1 2 - 3 252 84
NM_001145855.1 NP_001139327.1 2 - 3 395 84
NM_001145856.1 NP_001139328.1 2 - 3 342 84
NM_003023.4 NP_003014.3 2 - 3 393 84
GenBank mRNA AB000462.1 - 2 - 3 393 84
AB000463.1 - 2 - 3 393 84
AF000936.1 - 1 - 2 84
AK225497.1 - 2 - 3 254 84
AK299996.1 - 2 - 3 311 84
AK304752.1 - 2 - 3 297 84
AK307106.1 - 3 - 4 270 84
AK312286.1 - 2 - 3 190 84
BC022996.1 - 2 - 3 248 84
DC391267.1 - 2 - 3 396 84
U56386.1 - 2 - 3 340 84

View Full Product Details