Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRADD
Entrez Gene ID
8717
Gene Name
TNFRSF1A associated via death domain
Gene Aliases
Hs.89862
Chromosome Location
Chr.16: 67154185 - 67159909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323552.1 NP_001310481.1 2 - 3 247 91
NM_003789.3 NP_003780.1 2 - 3 234 91
XM_017023815.1 XP_016879304.1 2 - 3 300 91
GenBank mRNA AK315654.1 - 2 - 3 627 91
BC004491.2 - 2 - 3 211 91
BG717460.1 - 2 - 3 229 91
BT006934.1 - 1 - 2 154 91
L41690.1 - 2 - 3 202 91

View Full Product Details