Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHBDL1
Entrez Gene ID
9028
Gene Name
rhomboid like 1
Gene Aliases
RHBDL, RRP
Chromosome Location
Chr.16: 675666 - 678271 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278720.1 NP_001265649.1 3 - 4 552 54
NM_001318733.1 NP_001305662.1 2 - 3 649 54
XM_017023849.1 XP_016879338.1 2 - 3 652 54
XM_017023850.1 XP_016879339.1 2 - 3 652 54
GenBank mRNA AJ272344.1 - 3 - 4 512 54
AK294136.1 - 3 - 4 496 54
BC120874.2 - 2 - 3 641 54
BC120875.2 - 2 - 3 642 54
BI600093.1 - 3 - 4 560 54
Y17108.1 - 2 - 3 649 54

View Full Product Details