The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199279.1 | NP_001186208.1 | 1 - 2 | 439 | 68 |
NM_004198.3 | NP_004189.1 | 1 - 2 | 439 | 68 | |
GenBank mRNA | AK298798.1 | - | 1 - 2 | 263 | 68 |
AK313521.1 | - | 1 - 2 | 285 | 68 | |
BC014456.1 | - | 1 - 2 | 129 | 68 | |
DC307404.1 | - | 1 - 2 | 441 | 68 | |
U62435.1 | - | 1 - 2 | 225 | 68 | |
Y16282.1 | - | 1 - 2 | 83 | 68 |