The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303422.1 | NP_001290351.1 | 7 - 8 | 939 | 80 |
NM_004490.2 | NP_004481.2 | 8 - 9 | 1568 | 80 | |
XM_017003899.1 | XP_016859388.1 | 8 - 9 | 1005 | 80 | |
GenBank mRNA | AK301961.1 | - | 7 - 8 | 884 | 80 |
AK302129.1 | - | 6 - 7 | 1087 | 80 | |
AK307744.1 | - | 8 - 9 | 1081 | 80 | |
AK313153.1 | - | 8 - 9 | 1197 | 80 | |
BC053559.1 | - | 8 - 9 | 1038 | 80 | |
L76687.1 | - | 8 - 9 | 1567 | 80 |