Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HGS
Entrez Gene ID
9146
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Gene Aliases
HRS
Chromosome Location
Chr.17: 81683932 - 81702121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004712.4 NP_004703.1 21 - 22 2359 107
XM_017025297.1 XP_016880786.1 22 - 23 2516 107
XM_017025298.1 XP_016880787.1 20 - 21 2239 107
GenBank mRNA AF260566.1 - 20 - 21 1967 107
AK303800.1 - 19 - 20 1863 107
BC003565.1 - 21 - 22 2301 107
BT009754.1 - 21 - 22 2224 107
D84064.1 - 21 - 22 2284 107
KU178466.1 - 21 - 22 2224 107
KU178467.1 - 18 - 19 1990 107
U43895.1 - 21 - 22 2299 107

View Full Product Details