The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005049.2 | NP_005040.2 | 4 - 5 | 415 | 64 |
XM_011529667.2 | XP_011527969.1 | 4 - 5 | 480 | 64 | |
GenBank mRNA | AK314184.1 | - | 4 - 5 | 419 | 64 |
BC013309.2 | - | 4 - 5 | 325 | 64 | |
BC014988.1 | - | 4 - 5 | 325 | 64 | |
U53346.1 | - | 4 - 5 | 349 | 64 | |
U56085.1 | - | 4 - 5 | 360 | 64 | |
X95263.1 | - | 4 - 5 | 356 | 64 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006723910.1 | XP_006723973.1 | 4 - 5 | 452 | 64 |