Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr.21: 44107262 - 44131181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005049.2 NP_005040.2 6 - 7 696 81
XM_011529667.2 XP_011527969.1 6 - 7 761 81
GenBank mRNA AK314184.1 - 6 - 7 700 81
BC013309.2 - 6 - 7 606 81
BC014988.1 - 6 - 7 606 81
U53346.1 - 6 - 7 630 81
U56085.1 - 6 - 7 641 81
X95263.1 - 6 - 7 637 81

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
-
Chromosome Location
Chr.21: 5130871 - 5154767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723910.1 XP_006723973.1 6 - 7 733 81

View Full Product Details