Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NET1
Entrez Gene ID
10276
Gene Name
neuroepithelial cell transforming 1
Gene Aliases
ARHGEF8, NET1A
Chromosome Location
Chr.10: 5412551 - 5459056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047160.2 NP_001040625.1 7 - 8 828 98
NM_005863.4 NP_005854.2 5 - 6 739 98
NR_073040.1 5 - 6 772 98
GenBank mRNA AB209852.1 - 7 - 8 774 98
AJ010046.1 - 5 - 6 672 98
AK024919.1 - 5 - 6 679 98
AK299335.1 - 7 - 8 784 98
AK302687.1 - 7 - 8 610 98
AK304028.1 - 5 - 6 714 98
AK315566.1 - 5 - 6 681 98
BC010285.1 - 5 - 6 670 98
BC053553.1 - 7 - 8 780 98
BX537509.1 - 2 - 3 2952 98
U02081.1 - 3 - 4 260 98

View Full Product Details