The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006403.3 | NP_006394.1 | 1 - 2 | 172 | 94 |
NM_182966.3 | NP_892011.2 | 1 - 2 | 172 | 94 | |
GenBank mRNA | BC020686.1 | - | 1 - 2 | 143 | 94 |
BC040207.1 | - | 1 - 2 | 158 | 94 | |
DA778416.1 | - | 1 - 2 | 172 | 94 | |
L43821.1 | - | 1 - 2 | 168 | 94 | |
U64317.1 | - | 1 - 2 | 143 | 94 |