Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TACC2
Entrez Gene ID
10579
Gene Name
transforming acidic coiled-coil containing protein 2
Gene Aliases
AZU-1, ECTACC
Chromosome Location
Chr.10: 121989174 - 122254545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291876.1 NP_001278805.1 17 - 18 8554 94
NM_001291877.1 NP_001278806.1 17 - 18 8701 94
NM_001291878.1 NP_001278807.1 14 - 15 3239 94
NM_001291879.1 NP_001278808.1 14 - 15 2912 94
NM_006997.3 NP_008928.1 14 - 15 3293 94
NM_206860.2 NP_996742.1 13 - 14 3203 94
NM_206861.2 NP_996743.1 17 - 18 3358 94
NM_206862.3 NP_996744.3 20 - 21 8920 94
XM_005269388.1 XP_005269445.1 21 - 22 8837 94
XM_005269389.4 XP_005269446.1 21 - 22 9361 94
XM_005269390.1 XP_005269447.1 22 - 23 8780 94
XM_005269391.1 XP_005269448.1 20 - 21 8777 94
XM_005269392.1 XP_005269449.1 20 - 21 8747 94
XM_005269393.1 XP_005269450.1 21 - 22 8744 94
XM_005269394.1 XP_005269451.1 20 - 21 8711 94
XM_005269395.1 XP_005269452.1 20 - 21 8702 94
XM_005269396.1 XP_005269453.1 19 - 20 8606 94
XM_005269397.1 XP_005269454.1 19 - 20 8576 94
XM_005269399.1 XP_005269456.1 18 - 19 8378 94
XM_006717548.1 XP_006717611.1 21 - 22 8732 94
XM_006717552.1 XP_006717615.1 20 - 21 8549 94
XM_006717553.1 XP_006717616.1 19 - 20 8513 94
XM_006717554.1 XP_006717617.1 19 - 20 8501 94
XM_011539126.1 XP_011537428.1 21 - 22 8834 94
XM_011539128.1 XP_011537430.1 19 - 20 3410 94
XM_011539129.1 XP_011537431.1 18 - 19 3275 94
XM_011539130.1 XP_011537432.1 18 - 19 3182 94
XM_011539131.1 XP_011537433.1 14 - 15 3248 94
XM_011539132.1 XP_011537434.1 15 - 16 3191 94
XM_011539133.1 XP_011537435.1 13 - 14 2865 94
XM_011539134.1 XP_011537436.1 13 - 14 3136 94
XM_017015484.1 XP_016870973.1 21 - 22 8825 94
XM_017015485.1 XP_016870974.1 21 - 22 8690 94
XM_017015486.1 XP_016870975.1 20 - 21 8654 94
XM_017015487.1 XP_016870976.1 20 - 21 8642 94
XM_017015488.1 XP_016870977.1 19 - 20 8471 94
XM_017015489.1 XP_016870978.1 18 - 19 8393 94
XM_017015491.1 XP_016870980.1 14 - 15 3388 94
XM_017015492.1 XP_016870981.1 14 - 15 3295 94
XM_017015493.1 XP_016870982.1 13 - 14 3053 94
XM_017015494.1 XP_016870983.1 12 - 13 3064 94
XM_017015495.1 XP_016870984.1 12 - 13 2924 94
XM_017015496.1 XP_016870985.1 15 - 16 3189 94
XM_017015497.1 XP_016870986.1 15 - 16 3448 94
XM_017015498.1 XP_016870987.1 14 - 15 3313 94
XM_017015499.1 XP_016870988.1 16 - 17 3282 94
GenBank mRNA AF095791.2 - 14 - 15 2880 94
AF176646.1 - 13 - 14 3038 94
AF220152.2 - 10 - 11 2726 94
AF528098.1 - 17 - 18 3339 94
AF528099.1 - 20 - 21 8901 94
AK027066.1 - 10 - 11 1832 94
AK056743.1 - 1 - 2 1149 94
AK075097.1 - 10 - 11 1190 94
AK095371.1 - 1 - 2 2338 94
AL713712.1 - 10 - 11 2138 94
AL833304.1 - 19 - 20 8019 94
BC000999.2 - 3 - 4 287 94
BC039311.1 - 13 - 14 3203 94
BC141837.1 - 15 - 16 2891 94
BC142673.1 - 15 - 16 2891 94
BC144599.1 - 17 - 18 8669 94
BC144600.1 - 17 - 18 8522 94
BC150627.1 - 17 - 18 8522 94

View Full Product Details