Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC2
Entrez Gene ID
10579
Gene Name
transforming acidic coiled-coil containing protein 2
Gene Aliases
AZU-1, ECTACC
Chromosome Location
Chr.10: 121989174 - 122254545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291878.1 NP_001278807.1 9 - 10 2672 60
NM_006997.3 NP_008928.1 8 - 9 2636 60
NM_206860.2 NP_996742.1 8 - 9 2636 60
NM_206861.2 NP_996743.1 11 - 12 2701 60
NM_206862.3 NP_996744.3 14 - 15 8263 60
XM_005269388.1 XP_005269445.1 15 - 16 8180 60
XM_005269389.4 XP_005269446.1 15 - 16 8704 60
XM_005269390.1 XP_005269447.1 16 - 17 8123 60
XM_005269391.1 XP_005269448.1 14 - 15 8120 60
XM_005269392.1 XP_005269449.1 15 - 16 8180 60
XM_005269393.1 XP_005269450.1 15 - 16 8087 60
XM_005269394.1 XP_005269451.1 14 - 15 8054 60
XM_005269395.1 XP_005269452.1 14 - 15 8045 60
XM_005269397.1 XP_005269454.1 13 - 14 7919 60
XM_006717548.1 XP_006717611.1 15 - 16 8075 60
XM_011539126.1 XP_011537428.1 15 - 16 8177 60
XM_011539128.1 XP_011537430.1 13 - 14 2753 60
XM_011539129.1 XP_011537431.1 12 - 13 2618 60
XM_011539130.1 XP_011537432.1 12 - 13 2525 60
XM_011539131.1 XP_011537433.1 8 - 9 2591 60
XM_011539132.1 XP_011537434.1 9 - 10 2534 60
XM_017015484.1 XP_016870973.1 15 - 16 8168 60
XM_017015485.1 XP_016870974.1 16 - 17 8123 60
XM_017015486.1 XP_016870975.1 15 - 16 8087 60
XM_017015487.1 XP_016870976.1 15 - 16 8075 60
XM_017015488.1 XP_016870977.1 13 - 14 7814 60
XM_017015489.1 XP_016870978.1 13 - 14 7826 60
XM_017015491.1 XP_016870980.1 8 - 9 2731 60
XM_017015492.1 XP_016870981.1 8 - 9 2638 60
XM_017015493.1 XP_016870982.1 8 - 9 2486 60
XM_017015496.1 XP_016870985.1 9 - 10 2532 60
XM_017015497.1 XP_016870986.1 9 - 10 2791 60
XM_017015498.1 XP_016870987.1 8 - 9 2656 60
XM_017015499.1 XP_016870988.1 11 - 12 2715 60
GenBank mRNA AF095791.2 - 8 - 9 2223 60
AF528098.1 - 11 - 12 2682 60
AF528099.1 - 14 - 15 8244 60
AL833304.1 - 13 - 14 7362 60
BC039311.1 - 8 - 9 2636 60
BC141837.1 - 10 - 11 2324 60
BC142673.1 - 10 - 11 2324 60

View Full Product Details