Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBTF
Entrez Gene ID
7343
Gene Name
upstream binding transcription factor, RNA polymerase I
Gene Aliases
NOR-90, UBF, UBF-1, UBF1, UBF2
Chromosome Location
Chr.17: 44205033 - 44221626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014233.3 NP_055048.1 7 - 8 963 73
NR_045058.1 7 - 8 1160 73
XM_006722059.3 XP_006722122.1 7 - 8 1002 73
XM_006722060.2 XP_006722123.1 6 - 7 746 73
XM_006722061.2 XP_006722124.1 7 - 8 830 73
XM_017024995.1 XP_016880484.1 7 - 8 1011 73
XM_017024996.1 XP_016880485.1 7 - 8 963 73
XM_017024997.1 XP_016880486.1 7 - 8 1155 73
XM_017024998.1 XP_016880487.1 7 - 8 871 73
XM_017024999.1 XP_016880488.1 6 - 7 746 73
GenBank mRNA AK291733.1 - 7 - 8 788 73
AK292518.1 - 7 - 8 1160 73
AK292577.1 - 7 - 8 1048 73
X53390.1 - 6 - 7 814 73
X53461.1 - 6 - 7 814 73

View Full Product Details