The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014243.2 | NP_055058.2 | 10 - 11 | 1524 | 124 |
XM_011532421.2 | XP_011530723.1 | 10 - 11 | 1556 | 124 | |
XM_011532422.2 | XP_011530724.1 | 10 - 11 | 2055 | 124 | |
GenBank mRNA | AB002364.1 | - | 10 - 11 | 1476 | 124 |
BC130287.1 | - | 10 - 11 | 1521 | 124 | |
BC132735.1 | - | 10 - 11 | 1521 | 124 |