The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171166.1 | NP_001164637.1 | 7 - 8 | 745 | 64 |
NM_001171167.1 | NP_001164638.1 | 8 - 9 | 1184 | 64 | |
NM_001171168.1 | NP_001164639.1 | 7 - 8 | 1112 | 64 | |
NM_015099.3 | NP_055914.2 | 8 - 9 | 840 | 64 | |
XM_006721478.3 | XP_006721541.1 | 8 - 9 | 1487 | 64 | |
XM_006721481.3 | XP_006721544.1 | 7 - 8 | 1400 | 64 | |
XM_006721482.3 | XP_006721545.1 | 6 - 7 | 533 | 64 | |
XM_011523746.2 | XP_011522048.1 | 8 - 9 | 847 | 64 | |
XM_011523747.2 | XP_011522049.1 | 8 - 9 | 831 | 64 | |
XM_011523748.2 | XP_011522050.1 | 8 - 9 | 846 | 64 | |
XM_011523749.2 | XP_011522051.1 | 8 - 9 | 830 | 64 | |
GenBank mRNA | AB020716.1 | - | 8 - 9 | 801 | 64 |
AK302528.1 | - | 8 - 9 | 779 | 64 | |
AL831849.1 | - | 7 - 8 | 1112 | 64 | |
AL833974.1 | - | 7 - 8 | 730 | 64 | |
BC136534.1 | - | 7 - 8 | 769 | 64 | |
BC142695.1 | - | 8 - 9 | 831 | 64 | |
BC144232.1 | - | 6 - 7 | 697 | 64 | |
BC167148.1 | - | 8 - 9 | 801 | 64 | |
BC167149.1 | - | 8 - 9 | 801 | 64 | |
BC167151.1 | - | 8 - 9 | 801 | 64 | |
BC167160.1 | - | 8 - 9 | 801 | 64 |