Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSX5
Entrez Gene ID
6758
Gene Name
SSX family member 5
Gene Aliases
-
Chromosome Location
Chr.X: 48177914 - 48196798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021015.3 NP_066295.3 2 - 3 126 70
GenBank mRNA BC016640.2 - 1 - 2 92 70
BE896093.1 - 1 - 2 106 70

View Full Product Details