The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256336.1 | NP_001243265.1 | 20 - 21 | 2462 | 87 |
NM_001256337.1 | NP_001243266.1 | 16 - 17 | 2039 | 87 | |
NM_001256338.1 | NP_001243267.1 | 15 - 16 | 1957 | 87 | |
NM_004284.4 | NP_004275.4 | 21 - 22 | 2569 | 87 | |
NM_024568.2 | NP_078844.2 | 21 - 22 | 2541 | 87 | |
NR_046070.1 | 19 - 20 | 2384 | 87 | ||
XM_006711639.3 | XP_006711702.1 | 20 - 21 | 2437 | 87 | |
XM_017002858.1 | XP_016858347.1 | 22 - 23 | 3297 | 87 | |
XM_017002859.1 | XP_016858348.1 | 21 - 22 | 2424 | 87 | |
XM_017002860.1 | XP_016858349.1 | 21 - 22 | 2510 | 87 | |
XM_017002861.1 | XP_016858350.1 | 20 - 21 | 2443 | 87 | |
XM_017002862.1 | XP_016858351.1 | 13 - 14 | 2042 | 87 | |
XM_017002863.1 | XP_016858352.1 | 19 - 20 | 2258 | 87 | |
XM_017002864.1 | XP_016858353.1 | 20 - 21 | 2436 | 87 | |
GenBank mRNA | AF537213.1 | - | 21 - 22 | 2526 | 87 |
AK026183.1 | - | 9 - 10 | 1221 | 87 | |
AK027631.1 | - | 21 - 22 | 2497 | 87 | |
AK293157.1 | - | 16 - 17 | 1974 | 87 | |
AK297135.1 | - | 15 - 16 | 1895 | 87 | |
AK298424.1 | - | 20 - 21 | 2398 | 87 | |
BC001171.1 | - | 21 - 22 | 2503 | 87 | |
BC005038.1 | - | 18 - 19 | 2184 | 87 | |
BC008649.1 | - | 7 - 8 | 976 | 87 | |
BC043501.1 | - | 21 - 22 | 2487 | 87 | |
BC077717.1 | - | 15 - 16 | 1894 | 87 | |
BG195857.1 | - | 1 - 2 | 119 | 87 | |
EF560738.1 | - | 21 - 22 | 2537 | 87 | |
KJ534809.1 | - | 21 - 22 | 2506 | 87 |