The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133265.2 | NP_573572.1 | 1 - 2 | 363 | 113 |
GenBank mRNA | AF286598.1 | - | 1 - 2 | 363 | 113 |
AK301573.1 | - | - | 372 | 113 |