Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
Chromosome Location
Chr.X: 112774877 - 112840946 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113490.1 NP_001106962.1 2 - 3 1390 67
NM_133265.2 NP_573572.1 3 - 4 959 67
XM_005262087.1 XP_005262144.1 5 - 6 2135 67
XM_005262090.1 XP_005262147.1 2 - 3 632 67
XM_011530875.2 XP_011529177.1 6 - 7 2247 67
XM_017029289.1 XP_016884778.1 4 - 5 1163 67
GenBank mRNA AB028994.3 - 1 - 2 163 67
AF286598.1 - 3 - 4 959 67
AK301573.1 - - 1626 67
AY987378.1 - 2 - 3 1390 67
BC094712.1 - 3 - 4 1465 67
BC130294.1 - 1 - 2 380 67

View Full Product Details