Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
Chromosome Location
Chr.X: 112774877 - 112840946 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113490.1 NP_001106962.1 4 - 5 1630 83
NM_133265.2 NP_573572.1 5 - 6 1199 83
XM_005262087.1 XP_005262144.1 7 - 8 2375 83
XM_005262090.1 XP_005262147.1 4 - 5 872 83
XM_011530875.2 XP_011529177.1 8 - 9 2487 83
XM_017029289.1 XP_016884778.1 6 - 7 1403 83
GenBank mRNA AB028994.3 - 3 - 4 403 83
AF286598.1 - 5 - 6 1199 83
AY987378.1 - 4 - 5 1630 83
BC130294.1 - 3 - 4 620 83

View Full Product Details