Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEK10
Entrez Gene ID
152110
Gene Name
NIMA related kinase 10
Gene Aliases
-
Chromosome Location
Chr.3: 27107336 - 27369460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031741.3 NP_001026911.1 9 - 10 1083 66
NM_001304384.1 NP_001291313.1 9 - 10 1083 66
NM_152534.4 NP_689747.3 31 - 32 3183 66
XM_006712997.3 XP_006713060.1 31 - 32 3144 66
XM_006712998.2 XP_006713061.1 31 - 32 3144 66
XM_006712999.3 XP_006713062.1 31 - 32 3144 66
XM_017005761.1 XP_016861250.1 30 - 31 3096 66
XM_017005762.1 XP_016861251.1 32 - 33 3260 66
XM_017005763.1 XP_016861252.1 31 - 32 3241 66
XM_017005764.1 XP_016861253.1 30 - 31 3073 66
XM_017005765.1 XP_016861254.1 30 - 31 3045 66
XM_017005766.1 XP_016861255.1 30 - 31 3096 66
XM_017005767.1 XP_016861256.1 29 - 30 3009 66
XM_017005768.1 XP_016861257.1 30 - 31 3006 66
GenBank mRNA AK057247.1 - 9 - 10 1035 66
AK098832.1 - 12 - 13 1266 66
BC045758.1 - 9 - 10 1035 66

View Full Product Details