Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
Chromosome Location
Chr.15: 49326997 - 49620931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152647.2 NP_689860.2 3 - 4 396 106
XM_005254213.3 XP_005254270.1 3 - 4 473 106
XM_005254214.3 XP_005254271.1 3 - 4 406 106
XM_005254215.3 XP_005254272.1 3 - 4 473 106
XM_005254216.3 XP_005254273.1 3 - 4 473 106
XM_005254219.3 XP_005254276.1 3 - 4 473 106
XM_005254220.3 XP_005254277.1 3 - 4 473 106
XM_006720423.2 XP_006720486.1 4 - 5 438 106
XM_006720424.2 XP_006720487.1 3 - 4 473 106
XM_006720426.2 XP_006720489.1 3 - 4 473 106
XM_011521319.2 XP_011519621.1 3 - 4 2356 106
XM_011521321.2 XP_011519623.1 3 - 4 473 106
XM_011521324.1 XP_011519626.1 3 - 4 473 106
XM_011521325.2 XP_011519627.1 3 - 4 473 106
XM_011521326.1 XP_011519628.1 3 - 4 473 106
XM_017021990.1 XP_016877479.1 3 - 4 473 106
XM_017021993.1 XP_016877482.1 3 - 4 473 106
XM_017021994.1 XP_016877483.1 3 - 4 473 106
XM_017021995.1 XP_016877484.1 3 - 4 473 106
GenBank mRNA AK057362.1 - 3 - 4 394 106
BC048125.1 - 3 - 4 398 106
BC048139.1 - 3 - 4 398 106

View Full Product Details