Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPP7
Entrez Gene ID
143098
Gene Name
membrane palmitoylated protein 7
Gene Aliases
-
Chromosome Location
Chr.10: 28050993 - 28303066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318170.1 NP_001305099.1 10 - 11 1226 79
NM_173496.4 NP_775767.2 12 - 13 1252 79
NR_134517.1 10 - 11 1226 79
NR_134518.1 9 - 10 1107 79
XM_005252369.3 XP_005252426.1 10 - 11 1184 79
XM_011519337.2 XP_011517639.1 11 - 12 1151 79
XM_011519338.2 XP_011517640.1 9 - 10 1107 79
XM_017015741.1 XP_016871230.1 9 - 10 1864 79
XM_017015742.1 XP_016871231.1 10 - 11 1165 79
XM_017015743.1 XP_016871232.1 9 - 10 1868 79
GenBank mRNA AK057360.2 - 10 - 11 1149 79
AK130972.1 - 2 - 3 281 79
AK301592.1 - 9 - 10 1107 79
AK315046.1 - 10 - 11 1171 79
AL832380.1 - 9 - 10 1022 79
BC038105.2 - 12 - 13 1244 79

View Full Product Details