Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEL1L3
Entrez Gene ID
23231
Gene Name
SEL1L family member 3
Gene Aliases
Sel-1L3
Chromosome Location
Chr.4: 25697790 - 25863595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297592.1 NP_001284521.1 18 - 19 2740 99
NM_001297594.1 NP_001284523.1 18 - 19 2762 99
NM_015187.4 NP_056002.2 18 - 19 2957 99
XM_011513819.2 XP_011512121.2 18 - 19 2951 99
GenBank mRNA AB018289.1 - 17 - 18 2455 99
AK025282.1 - 6 - 7 623 99
AK289687.1 - 18 - 19 2705 99
AK300219.1 - 9 - 10 1055 99
BC009945.2 - 11 - 12 1472 99
BC041046.1 - 9 - 10 1109 99
BC060867.1 - 18 - 19 2750 99
CR749817.1 - 18 - 19 2740 99

View Full Product Details