The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297592.1 | NP_001284521.1 | 10 - 11 | 1752 | 93 |
NM_001297594.1 | NP_001284523.1 | 10 - 11 | 1774 | 93 | |
NM_015187.4 | NP_056002.2 | 10 - 11 | 1969 | 93 | |
XM_011513819.2 | XP_011512121.2 | 10 - 11 | 1963 | 93 | |
XM_017007923.1 | XP_016863412.1 | 9 - 10 | 1607 | 93 | |
GenBank mRNA | AB018289.1 | - | 9 - 10 | 1467 | 93 |
AK289687.1 | - | 10 - 11 | 1717 | 93 | |
AK300219.1 | - | 1 - 2 | 67 | 93 | |
BC009945.2 | - | 3 - 4 | 484 | 93 | |
BC041046.1 | - | 1 - 2 | 121 | 93 | |
BC060867.1 | - | 10 - 11 | 1762 | 93 | |
CR749817.1 | - | 10 - 11 | 1752 | 93 |