The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042406.1 | NP_001035865.1 | 5 - 6 | 700 | 72 |
NM_001287741.1 | NP_001274670.1 | 4 - 5 | 604 | 72 | |
NM_001287746.1 | NP_001274675.1 | 6 - 7 | 796 | 72 | |
NM_019036.2 | NP_061909.2 | 6 - 7 | 790 | 72 | |
XM_011514700.2 | XP_011513002.1 | 5 - 6 | 700 | 72 | |
GenBank mRNA | AF131827.1 | - | 1 - 2 | 94 | 72 |
AK055075.1 | - | 6 - 7 | 681 | 72 | |
AK127587.1 | - | 6 - 7 | 763 | 72 | |
AK293856.1 | - | 4 - 5 | 465 | 72 | |
BC024194.2 | - | 5 - 6 | 581 | 72 |