The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017553.2 | NP_060023.1 | 27 - 28 | 3718 | 70 |
XM_011521685.2 | XP_011519987.1 | 27 - 28 | 4553 | 70 | |
XM_011521686.2 | XP_011519988.1 | 13 - 14 | 1529 | 70 | |
GenBank mRNA | AB033085.1 | - | 27 - 28 | 3614 | 70 |
AK026050.1 | - | 8 - 9 | 1068 | 70 | |
AK057582.1 | - | 16 - 17 | 1983 | 70 | |
AK290371.1 | - | 26 - 27 | 3456 | 70 | |
BC146785.1 | - | 27 - 28 | 3626 | 70 |