Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PPP4R1L
- Entrez Gene ID
-
55370
- Gene Name
- protein phosphatase 4 regulatory subunit 1 like (pseudogene)
- Gene Aliases
-
C20orf192, PPP4R1P1, PRO1085
- Chromosome Location
-
Chr.20: 58232777 - 58309439 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003505.3 |
|
13 - 14 |
1930 |
129 |
GenBank mRNA |
AK057583.1 |
- |
13 - 14 |
1930 |
129 |
CR994964.1 |
- |
3 - 4 |
380 |
129 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD654184.1 |
- |
2 - 3 |
274 |
|
Chr. 20 - 58232793 - 58239773 [-] on Build GRCh38 |
129 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ankyrin repeat domain 60 |
ANKRD60 |
C20orf86, bA196N14.3 |
-9.3kb
|
Sense |
Chr. - 58216126 - 58228653 on Build GRCh38 |
Downstream |
RAB22A, member RAS oncogene family |
RAB22A |
|
70.3kb
|
Anti-sense |
Chr. - 58309715 - 58367507 on Build GRCh38 |
View Full Product Details