The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242581.1 | NP_001229510.1 | 5 - 6 | 755 | 69 |
NM_018993.3 | NP_061866.1 | 4 - 5 | 495 | 69 | |
XM_005260731.2 | XP_005260788.1 | 6 - 7 | 951 | 69 | |
XM_006723574.3 | XP_006723637.1 | 7 - 8 | 933 | 69 | |
XM_006723575.3 | XP_006723638.1 | 6 - 7 | 719 | 69 | |
XM_006723577.2 | XP_006723640.1 | 5 - 6 | 568 | 69 | |
XM_011529255.2 | XP_011527557.2 | 5 - 6 | 1028 | 69 | |
XM_011529257.1 | XP_011527559.1 | 5 - 6 | 624 | 69 | |
XM_011529258.2 | XP_011527560.1 | 5 - 6 | 636 | 69 | |
XM_011529259.2 | XP_011527561.1 | 4 - 5 | 548 | 69 | |
XM_017027887.1 | XP_016883376.1 | 8 - 9 | 1274 | 69 | |
XM_017027888.1 | XP_016883377.1 | 9 - 10 | 1346 | 69 | |
XM_017027889.1 | XP_016883378.1 | 4 - 5 | 875 | 69 | |
XM_017027890.1 | XP_016883379.1 | 6 - 7 | 706 | 69 | |
XM_017027891.1 | XP_016883380.1 | 6 - 7 | 744 | 69 | |
XM_017027892.1 | XP_016883381.1 | 4 - 5 | 559 | 69 | |
XM_017027893.1 | XP_016883382.1 | 5 - 6 | 1053 | 69 | |
GenBank mRNA | AK094884.1 | - | 5 - 6 | 756 | 69 |
AL136924.1 | - | 3 - 4 | 376 | 69 |