Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2595654-2595927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
UniGene
Hs.459691
Gene Chromosome Location
Chr.16: 2537964-2603190 on Build GRCh38
Transcripts
NM_001261816.1 NM_002613.4 NM_031268.5 XM_011522521.2 XM_011522523.2 XM_017023271.1 XM_017023272.1 XM_017023273.1 XM_017023274.1 XM_017023275.1 XM_017023276.1 XM_017023277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4713400 c.1383C>T p.G461G Chr.16: 2595832-2595832 on Build GRCh38
5609815 c.1390G>A p.D464N Chr.16: 2595839-2595839 on Build GRCh38
6909069 c.1397G>A p.R466Q Chr.16: 2595846-2595846 on Build GRCh38
6978012 c.1344G>T p.W448C Chr.16: 2595793-2595793 on Build GRCh38
3420901 c.1396C>T p.R466W Chr.16: 2595845-2595845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779859162 Chr.16: 2595869-2595869 on Build GRCh38
rs752063594 Chr.16: 2595789-2595789 on Build GRCh38
rs776699427 Chr.16: 2595750-2595750 on Build GRCh38
rs768597146 Chr.16: 2595873-2595873 on Build GRCh38
rs369196937 Chr.16: 2595901-2595901 on Build GRCh38
rs968691533 Chr.16: 2595717-2595717 on Build GRCh38
rs371663580 Chr.16: 2595867-2595867 on Build GRCh38
rs202230134 Chr.16: 2595881-2595881 on Build GRCh38
rs546784059 Chr.16: 2595692-2595692 on Build GRCh38
rs763256086 Chr.16: 2595882-2595882 on Build GRCh38
rs777350674 Chr.16: 2595815-2595815 on Build GRCh38
rs762040074 Chr.16: 2595756-2595756 on Build GRCh38
rs368701059 Chr.16: 2595790-2595790 on Build GRCh38
rs960148628 Chr.16: 2595700-2595700 on Build GRCh38
rs764122924 Chr.16: 2595748-2595748 on Build GRCh38
rs932084836 Chr.16: 2595744-2595745 on Build GRCh38
rs372825770 Chr.16: 2595734-2595734 on Build GRCh38
rs760545916 Chr.16: 2595860-2595860 on Build GRCh38
rs746886761 Chr.16: 2595870-2595870 on Build GRCh38
rs569124094 Chr.16: 2595853-2595853 on Build GRCh38
rs139080760 Chr.16: 2595678-2595678 on Build GRCh38
rs770023184 Chr.16: 2595878-2595878 on Build GRCh38
rs942224864 Chr.16: 2595871-2595871 on Build GRCh38
rs776995460 Chr.16: 2595906-2595907 on Build GRCh38
rs200543006 Chr.16: 2595760-2595760 on Build GRCh38
rs753410336 Chr.16: 2595845-2595845 on Build GRCh38
rs150044605 Chr.16: 2595727-2595727 on Build GRCh38
rs374649760 Chr.16: 2595809-2595809 on Build GRCh38
rs757520748 Chr.16: 2595858-2595858 on Build GRCh38
rs548974990 Chr.16: 2595787-2595787 on Build GRCh38
rs766808966 Chr.16: 2595888-2595888 on Build GRCh38
rs929541153 Chr.16: 2595900-2595900 on Build GRCh38
rs566871879 Chr.16: 2595718-2595718 on Build GRCh38
rs916110147 Chr.16: 2595739-2595739 on Build GRCh38
rs200084489 Chr.16: 2595877-2595877 on Build GRCh38
rs376061888 Chr.16: 2595890-2595890 on Build GRCh38
rs745653878 Chr.16: 2595860-2595860 on Build GRCh38
rs760061971 Chr.16: 2595898-2595898 on Build GRCh38
rs765431759 Chr.16: 2595761-2595761 on Build GRCh38
rs374466740 Chr.16: 2595766-2595766 on Build GRCh38
rs1020122848 Chr.16: 2595701-2595701 on Build GRCh38
rs537325699 Chr.16: 2595757-2595757 on Build GRCh38

View Full Product Details