Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44793403-44793670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620916 c.1167C>T p.C389C Chr.19: 44793503-44793503 on Build GRCh38
6345643 c.1284+1G>T p.? Chr.19: 44793621-44793621 on Build GRCh38
7422023 c.1187G>T p.W396L Chr.19: 44793523-44793523 on Build GRCh38
3960196 c.1227T>A p.T409T Chr.19: 44793563-44793563 on Build GRCh38
5020654 c.1239A>G p.S413S Chr.19: 44793575-44793575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772940892 Chr.19: 44793450-44793450 on Build GRCh38
rs146735092 Chr.19: 44793495-44793495 on Build GRCh38
rs200336706 Chr.19: 44793461-44793461 on Build GRCh38
rs535354110 Chr.19: 44793479-44793479 on Build GRCh38
rs1043138391 Chr.19: 44793539-44793539 on Build GRCh38
rs745326746 Chr.19: 44793437-44793437 on Build GRCh38
rs994347051 Chr.19: 44793603-44793603 on Build GRCh38
rs3208856 Chr.19: 44793549-44793549 on Build GRCh38
rs869161737 Chr.19: 44793590-44793591 on Build GRCh38
rs775347030 Chr.19: 44793467-44793467 on Build GRCh38
rs370828809 Chr.19: 44793462-44793462 on Build GRCh38
rs765888811 Chr.19: 44793437-44793446 on Build GRCh38
rs61750955 Chr.19: 44793575-44793575 on Build GRCh38
rs753854663 Chr.19: 44793584-44793584 on Build GRCh38
rs374104325 Chr.19: 44793459-44793459 on Build GRCh38
rs77920506 Chr.19: 44793591-44793591 on Build GRCh38
rs201433561 Chr.19: 44793578-44793578 on Build GRCh38
rs751385301 Chr.19: 44793504-44793504 on Build GRCh38
rs768506085 Chr.19: 44793575-44793576 on Build GRCh38
rs754761911 Chr.19: 44793505-44793505 on Build GRCh38
rs746979867 Chr.19: 44793439-44793439 on Build GRCh38
rs572998027 Chr.19: 44793421-44793421 on Build GRCh38
rs374326666 Chr.19: 44793502-44793502 on Build GRCh38
rs772388756 Chr.19: 44793558-44793558 on Build GRCh38
rs374717459 Chr.19: 44793635-44793635 on Build GRCh38
rs762183942 Chr.19: 44793588-44793588 on Build GRCh38
rs754849774 Chr.19: 44793595-44793595 on Build GRCh38
rs112128460 Chr.19: 44793530-44793530 on Build GRCh38
rs758231610 Chr.19: 44793435-44793435 on Build GRCh38
rs769396781 Chr.19: 44793518-44793518 on Build GRCh38
rs749036591 Chr.19: 44793531-44793531 on Build GRCh38
rs768709265 Chr.19: 44793441-44793441 on Build GRCh38
rs17852620 Chr.19: 44793573-44793573 on Build GRCh38
rs118085256 Chr.19: 44793637-44793637 on Build GRCh38
rs756001540 Chr.19: 44793636-44793636 on Build GRCh38
rs748765725 Chr.19: 44793643-44793643 on Build GRCh38
rs765569181 Chr.19: 44793591-44793591 on Build GRCh38
rs750604085 Chr.19: 44793593-44793593 on Build GRCh38
rs757386289 Chr.19: 44793500-44793500 on Build GRCh38
rs368197310 Chr.19: 44793430-44793430 on Build GRCh38
rs753138447 Chr.19: 44793478-44793478 on Build GRCh38
rs560728785 Chr.19: 44793617-44793617 on Build GRCh38
rs368494441 Chr.19: 44793552-44793552 on Build GRCh38
rs762865111 Chr.19: 44793531-44793532 on Build GRCh38
rs781113299 Chr.19: 44793597-44793597 on Build GRCh38
rs35457630 Chr.19: 44793510-44793510 on Build GRCh38
rs950413191 Chr.19: 44793614-44793614 on Build GRCh38
rs764271266 Chr.19: 44793583-44793583 on Build GRCh38
rs1025909626 Chr.19: 44793452-44793452 on Build GRCh38
rs754137227 Chr.19: 44793497-44793497 on Build GRCh38
rs1025942028 Chr.19: 44793612-44793612 on Build GRCh38
rs747705475 Chr.19: 44793443-44793443 on Build GRCh38
rs769117489 Chr.19: 44793449-44793449 on Build GRCh38
rs777117772 Chr.19: 44793529-44793529 on Build GRCh38
rs1053116620 Chr.19: 44793516-44793516 on Build GRCh38
rs776338482 Chr.19: 44793442-44793442 on Build GRCh38
rs773939139 Chr.19: 44793551-44793551 on Build GRCh38
rs1025992203 Chr.19: 44793581-44793581 on Build GRCh38
rs66944506 Chr.19: 44793589-44793590 on Build GRCh38
rs138817753 Chr.19: 44793438-44793438 on Build GRCh38
rs200326363 Chr.19: 44793469-44793469 on Build GRCh38
rs781277300 Chr.19: 44793509-44793509 on Build GRCh38
rs543773390 Chr.19: 44793467-44793467 on Build GRCh38

View Full Product Details