Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44780768-44781041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998051 c.356G>A p.R119Q Chr.19: 44780907-44780907 on Build GRCh38
6384532 c.419C>T p.P140L Chr.19: 44780970-44780970 on Build GRCh38
5048328 c.393G>T p.M131I Chr.19: 44780944-44780944 on Build GRCh38
3971084 c.417C>T p.F139F Chr.19: 44780968-44780968 on Build GRCh38
6372983 c.482G>A p.R161K Chr.19: 44781033-44781033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008498538 Chr.19: 44780816-44780816 on Build GRCh38
rs750395712 Chr.19: 44780875-44780875 on Build GRCh38
rs765128314 Chr.19: 44780951-44780951 on Build GRCh38
rs747887583 Chr.19: 44780976-44780976 on Build GRCh38
rs757390586 Chr.19: 44781020-44781021 on Build GRCh38
rs369091435 Chr.19: 44780886-44780886 on Build GRCh38
rs140833100 Chr.19: 44780946-44780946 on Build GRCh38
rs374333860 Chr.19: 44780971-44780971 on Build GRCh38
rs764313148 Chr.19: 44780943-44780954 on Build GRCh38
rs981330867 Chr.19: 44780968-44780968 on Build GRCh38
rs199680893 Chr.19: 44780947-44780947 on Build GRCh38
rs935339144 Chr.19: 44780909-44780909 on Build GRCh38
rs775004600 Chr.19: 44781001-44781001 on Build GRCh38
rs757078394 Chr.19: 44780871-44780871 on Build GRCh38
rs770219520 Chr.19: 44780902-44780902 on Build GRCh38
rs891273833 Chr.19: 44780802-44780802 on Build GRCh38
rs751438592 Chr.19: 44780970-44780970 on Build GRCh38
rs371315315 Chr.19: 44780917-44780917 on Build GRCh38
rs752806081 Chr.19: 44780864-44780864 on Build GRCh38
rs2965127 Chr.19: 44780912-44780912 on Build GRCh38
rs775409459 Chr.19: 44781011-44781011 on Build GRCh38
rs375723461 Chr.19: 44780859-44780859 on Build GRCh38
rs779951141 Chr.19: 44780962-44780962 on Build GRCh38
rs144857168 Chr.19: 44780978-44780978 on Build GRCh38
rs760078494 Chr.19: 44781004-44781004 on Build GRCh38
rs747718914 Chr.19: 44780901-44780901 on Build GRCh38
rs755764885 Chr.19: 44780893-44780893 on Build GRCh38
rs767637879 Chr.19: 44780929-44780929 on Build GRCh38
rs768351418 Chr.19: 44780894-44780894 on Build GRCh38
rs35106910 Chr.19: 44781009-44781009 on Build GRCh38
rs749602901 Chr.19: 44780990-44780990 on Build GRCh38
rs142893738 Chr.19: 44780915-44780915 on Build GRCh38
rs377440313 Chr.19: 44780972-44780972 on Build GRCh38
rs751836061 Chr.19: 44780968-44780968 on Build GRCh38
rs771366662 Chr.19: 44780991-44780991 on Build GRCh38
rs775291844 Chr.19: 44780880-44780899 on Build GRCh38
rs376312238 Chr.19: 44780961-44780961 on Build GRCh38
rs372975672 Chr.19: 44780906-44780906 on Build GRCh38
rs150974051 Chr.19: 44780926-44780926 on Build GRCh38
rs371947989 Chr.19: 44780805-44780805 on Build GRCh38
rs199953740 Chr.19: 44780948-44780948 on Build GRCh38
rs750151047 Chr.19: 44780953-44780953 on Build GRCh38
rs914689240 Chr.19: 44780992-44780992 on Build GRCh38
rs774630684 Chr.19: 44780919-44780919 on Build GRCh38
rs762668366 Chr.19: 44780909-44780910 on Build GRCh38
rs539748936 Chr.19: 44780887-44780887 on Build GRCh38
rs1050333751 Chr.19: 44780798-44780798 on Build GRCh38
rs201551471 Chr.19: 44781016-44781016 on Build GRCh38
rs865916210 Chr.19: 44780969-44780969 on Build GRCh38
rs141019786 Chr.19: 44780959-44780959 on Build GRCh38
rs764214326 Chr.19: 44781017-44781017 on Build GRCh38
rs780832639 Chr.19: 44780897-44780897 on Build GRCh38
rs997055854 Chr.19: 44780793-44780793 on Build GRCh38
rs201563104 Chr.19: 44780907-44780907 on Build GRCh38
rs746515008 Chr.19: 44780888-44780888 on Build GRCh38
rs1015821828 Chr.19: 44780825-44780825 on Build GRCh38
rs778532554 Chr.19: 44780874-44780874 on Build GRCh38
rs758307910 Chr.19: 44780884-44780884 on Build GRCh38
rs373313232 Chr.19: 44780863-44780863 on Build GRCh38

View Full Product Details