Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186191937-186192174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280435 c.114G>T p.A38A Chr.4: 186191937-186191937 on Build GRCh38
4764165 c.140T>G p.M47R Chr.4: 186191963-186191963 on Build GRCh38
6387108 c.175C>G p.L59V Chr.4: 186191998-186191998 on Build GRCh38
6385915 c.171C>A p.Y57* Chr.4: 186191994-186191994 on Build GRCh38
4609057 c.198G>T p.M66I Chr.4: 186192021-186192021 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040774145 Chr.4: 186192097-186192097 on Build GRCh38
rs774390457 Chr.4: 186192067-186192067 on Build GRCh38
rs377425030 Chr.4: 186192037-186192037 on Build GRCh38
rs766465711 Chr.4: 186191980-186191980 on Build GRCh38
rs866494306 Chr.4: 186192092-186192092 on Build GRCh38
rs1016076116 Chr.4: 186192099-186192099 on Build GRCh38
rs950505004 Chr.4: 186192083-186192083 on Build GRCh38
rs894597248 Chr.4: 186192124-186192124 on Build GRCh38
rs369891403 Chr.4: 186192054-186192054 on Build GRCh38
rs370158024 Chr.4: 186192130-186192130 on Build GRCh38
rs376907847 Chr.4: 186191971-186191971 on Build GRCh38
rs762882864 Chr.4: 186191976-186191976 on Build GRCh38
rs931137718 Chr.4: 186192075-186192075 on Build GRCh38
rs768506789 Chr.4: 186192046-186192046 on Build GRCh38
rs906756906 Chr.4: 186192047-186192047 on Build GRCh38
rs576521079 Chr.4: 186192074-186192074 on Build GRCh38
rs374174110 Chr.4: 186191992-186191992 on Build GRCh38
rs562098490 Chr.4: 186192118-186192118 on Build GRCh38
rs562331817 Chr.4: 186192146-186192146 on Build GRCh38
rs893974930 Chr.4: 186192001-186192001 on Build GRCh38
rs756779672 Chr.4: 186192009-186192009 on Build GRCh38
rs768059118 Chr.4: 186191990-186191990 on Build GRCh38
rs933349743 Chr.4: 186191975-186191975 on Build GRCh38
rs369484576 Chr.4: 186191991-186191991 on Build GRCh38
rs745413794 Chr.4: 186192020-186192020 on Build GRCh38
rs119103285 Chr.4: 186192004-186192004 on Build GRCh38
rs763590093 Chr.4: 186191997-186191997 on Build GRCh38
rs974952244 Chr.4: 186192116-186192116 on Build GRCh38
rs553484886 Chr.4: 186192030-186192030 on Build GRCh38
rs532043235 Chr.4: 186192125-186192125 on Build GRCh38
rs758638577 Chr.4: 186192082-186192082 on Build GRCh38
rs756285586 Chr.4: 186192085-186192085 on Build GRCh38
rs756941465 Chr.4: 186192109-186192109 on Build GRCh38
rs946291316 Chr.4: 186192105-186192105 on Build GRCh38
rs911004473 Chr.4: 186192091-186192091 on Build GRCh38
rs746984953 Chr.4: 186192145-186192145 on Build GRCh38
rs80250369 Chr.4: 186192154-186192154 on Build GRCh38
rs760001831 Chr.4: 186191986-186191986 on Build GRCh38
rs778598903 Chr.4: 186192017-186192017 on Build GRCh38
rs11555701 Chr.4: 186192149-186192149 on Build GRCh38
rs200327267 Chr.4: 186192006-186192006 on Build GRCh38
rs780104160 Chr.4: 186192031-186192031 on Build GRCh38
rs760891257 Chr.4: 186192076-186192076 on Build GRCh38
rs769901483 Chr.4: 186191970-186191970 on Build GRCh38
rs767907319 Chr.4: 186192071-186192071 on Build GRCh38
rs764546798 Chr.4: 186192093-186192093 on Build GRCh38
rs545440191 Chr.4: 186192102-186192102 on Build GRCh38
rs781008069 Chr.4: 186192153-186192153 on Build GRCh38
rs1041966346 Chr.4: 186192108-186192108 on Build GRCh38
rs758390423 Chr.4: 186192028-186192028 on Build GRCh38

View Full Product Details