Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102954203-102954474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704498 c.541C>T p.P181S Chr.11: 102954252-102954252 on Build GRCh38
3442756 c.511+1G>A p.? Chr.11: 102954457-102954457 on Build GRCh38
6576964 c.522C>A p.D174E Chr.11: 102954271-102954271 on Build GRCh38
3868302 c.503G>A p.G168E Chr.11: 102954466-102954466 on Build GRCh38
922316 c.549C>T p.G183G Chr.11: 102954244-102954244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764647902 Chr.11: 102954254-102954254 on Build GRCh38
rs376889562 Chr.11: 102954265-102954265 on Build GRCh38
rs760029158 Chr.11: 102954322-102954322 on Build GRCh38
rs773111941 Chr.11: 102954298-102954298 on Build GRCh38
rs754927616 Chr.11: 102954285-102954285 on Build GRCh38
rs749832764 Chr.11: 102954445-102954445 on Build GRCh38
rs189379640 Chr.11: 102954274-102954274 on Build GRCh38
rs202231908 Chr.11: 102954281-102954281 on Build GRCh38
rs756865618 Chr.11: 102954251-102954251 on Build GRCh38
rs928951639 Chr.11: 102954278-102954278 on Build GRCh38
rs771020164 Chr.11: 102954314-102954314 on Build GRCh38
rs781151341 Chr.11: 102954289-102954289 on Build GRCh38
rs640198 Chr.11: 102954362-102954362 on Build GRCh38
rs761073692 Chr.11: 102954436-102954436 on Build GRCh38
rs761238796 Chr.11: 102954343-102954343 on Build GRCh38
rs767961495 Chr.11: 102954323-102954323 on Build GRCh38
rs906425989 Chr.11: 102954342-102954342 on Build GRCh38
rs749530867 Chr.11: 102954307-102954307 on Build GRCh38
rs34647629 Chr.11: 102954367-102954367 on Build GRCh38
rs1054877508 Chr.11: 102954429-102954429 on Build GRCh38
rs566922121 Chr.11: 102954358-102954358 on Build GRCh38
rs775773373 Chr.11: 102954277-102954277 on Build GRCh38
rs551944248 Chr.11: 102954375-102954375 on Build GRCh38
rs72987509 Chr.11: 102954435-102954435 on Build GRCh38
rs774880084 Chr.11: 102954319-102954319 on Build GRCh38
rs764817104 Chr.11: 102954444-102954444 on Build GRCh38
rs748076381 Chr.11: 102954291-102954291 on Build GRCh38
rs769978537 Chr.11: 102954294-102954294 on Build GRCh38
rs750351800 Chr.11: 102954425-102954425 on Build GRCh38
rs1000556506 Chr.11: 102954405-102954405 on Build GRCh38
rs1046499523 Chr.11: 102954334-102954334 on Build GRCh38

View Full Product Details