Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1363002-1363273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1 NM_001193388.3 NM_001193389.1 NM_001276414.1 XM_005255505.1 XM_011522610.1 XM_011522611.1 XM_011522612.1 XM_017023568.1 XM_017023569.1

Gene Symbol
UNKL
Entrez Gene ID
64718
Gene Name
unkempt family like zinc finger
Gene Aliases
C16orf28, ZC3H5L, ZC3HDC5L
UniGene
Hs.643536
Gene Chromosome Location
Chr.16: 1363205-1414749 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1 NM_001193388.3 NM_001193389.1 NM_001276414.1 XM_005255505.1 XM_011522610.1 XM_011522611.1 XM_011522612.1 XM_017023568.1 XM_017023569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2143664 c.874C>T p.P292S Chr.16: 1363047-1363047 on Build GRCh38
3377745 c.865G>A p.A289T Chr.16: 1363038-1363038 on Build GRCh38
391193 c.833_838delACTTGG p.N278_E280>K Chr.16: 1363006-1363011 on Build GRCh38
1301639 c.872C>T p.S291F Chr.16: 1363045-1363045 on Build GRCh38
5363768 c.905G>A p.R302H Chr.16: 1363078-1363078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762384924 Chr.16: 1363042-1363042 on Build GRCh38
rs758226003 Chr.16: 1363108-1363108 on Build GRCh38
rs751017692 Chr.16: 1363088-1363088 on Build GRCh38
rs7187001 Chr.16: 1363083-1363083 on Build GRCh38
rs570375445 Chr.16: 1363224-1363224 on Build GRCh38
rs189677035 Chr.16: 1363118-1363118 on Build GRCh38
rs60000415 Chr.16: 1363159-1363159 on Build GRCh38
rs376414184 Chr.16: 1363124-1363124 on Build GRCh38
rs752565624 Chr.16: 1363098-1363098 on Build GRCh38
rs1016284477 Chr.16: 1363189-1363189 on Build GRCh38
rs778955815 Chr.16: 1363097-1363097 on Build GRCh38
rs762046443 Chr.16: 1363082-1363082 on Build GRCh38
rs764577534 Chr.16: 1363037-1363037 on Build GRCh38
rs755243924 Chr.16: 1363095-1363095 on Build GRCh38
rs775991626 Chr.16: 1363077-1363077 on Build GRCh38
rs963753126 Chr.16: 1363192-1363192 on Build GRCh38
rs774790794 Chr.16: 1363079-1363079 on Build GRCh38
rs894916504 Chr.16: 1363197-1363197 on Build GRCh38
rs147955846 Chr.16: 1363046-1363046 on Build GRCh38
rs746913643 Chr.16: 1363073-1363073 on Build GRCh38
rs375719850 Chr.16: 1363121-1363121 on Build GRCh38
rs775705721 Chr.16: 1363045-1363046 on Build GRCh38
rs770600776 Chr.16: 1363031-1363031 on Build GRCh38
rs777517352 Chr.16: 1363109-1363109 on Build GRCh38
rs1021169186 Chr.16: 1363144-1363144 on Build GRCh38
rs761250229 Chr.16: 1363090-1363090 on Build GRCh38
rs938858489 Chr.16: 1363178-1363178 on Build GRCh38
rs375089519 Chr.16: 1363054-1363054 on Build GRCh38
rs769262505 Chr.16: 1363078-1363078 on Build GRCh38
rs149512696 Chr.16: 1363194-1363194 on Build GRCh38
rs377184531 Chr.16: 1363120-1363120 on Build GRCh38
rs772539062 Chr.16: 1363028-1363030 on Build GRCh38
rs928811901 Chr.16: 1363171-1363171 on Build GRCh38
rs901014889 Chr.16: 1363158-1363158 on Build GRCh38
rs761272543 Chr.16: 1363143-1363143 on Build GRCh38
rs774003749 Chr.16: 1363023-1363023 on Build GRCh38
rs747006896 Chr.16: 1363114-1363114 on Build GRCh38
rs893519932 Chr.16: 1363122-1363122 on Build GRCh38
rs570294976 Chr.16: 1363094-1363094 on Build GRCh38
rs374511482 Chr.16: 1363025-1363025 on Build GRCh38
rs561640998 Chr.16: 1363060-1363060 on Build GRCh38
rs141807632 Chr.16: 1363076-1363076 on Build GRCh38
rs754236475 Chr.16: 1363074-1363074 on Build GRCh38
rs886051685 Chr.16: 1363203-1363207 on Build GRCh38
rs193302860 Chr.16: 1363030-1363030 on Build GRCh38
rs778776422 Chr.16: 1363056-1363056 on Build GRCh38
rs775046997 Chr.16: 1363125-1363125 on Build GRCh38
rs778271120 Chr.16: 1363229-1363229 on Build GRCh38
rs768090398 Chr.16: 1363047-1363047 on Build GRCh38
rs755154799 Chr.16: 1363055-1363055 on Build GRCh38
rs150647450 Chr.16: 1363059-1363059 on Build GRCh38
rs1011942737 Chr.16: 1363052-1363052 on Build GRCh38
rs759088945 Chr.16: 1363036-1363036 on Build GRCh38
rs780260521 Chr.16: 1363053-1363053 on Build GRCh38
rs773712619 Chr.16: 1363135-1363135 on Build GRCh38
rs772624200 Chr.16: 1363129-1363129 on Build GRCh38
rs751864147 Chr.16: 1363041-1363041 on Build GRCh38
rs374497551 Chr.16: 1363119-1363119 on Build GRCh38
rs772723751 Chr.16: 1363026-1363026 on Build GRCh38
rs752191785 Chr.16: 1363234-1363234 on Build GRCh38
rs750563207 Chr.16: 1363048-1363048 on Build GRCh38
rs111882845 Chr.16: 1363187-1363187 on Build GRCh38
rs372062796 Chr.16: 1363051-1363051 on Build GRCh38
rs200471078 Chr.16: 1363035-1363035 on Build GRCh38

View Full Product Details