Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12648336-12648577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5388670 c.2500G>A p.G834R Chr.19: 12648336-12648336 on Build GRCh38
5056232 c.2477A>G p.E826G Chr.19: 12648359-12648359 on Build GRCh38
3403776 c.2445G>C p.R815S Chr.19: 12648391-12648391 on Build GRCh38
991364 c.2490G>T p.E830D Chr.19: 12648346-12648346 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371508056 Chr.19: 12648436-12648436 on Build GRCh38
rs767484276 Chr.19: 12648435-12648436 on Build GRCh38
rs1012498827 Chr.19: 12648552-12648552 on Build GRCh38
rs769427198 Chr.19: 12648405-12648405 on Build GRCh38
rs751431290 Chr.19: 12648428-12648429 on Build GRCh38
rs535248547 Chr.19: 12648533-12648533 on Build GRCh38
rs192419876 Chr.19: 12648505-12648505 on Build GRCh38
rs773223523 Chr.19: 12648534-12648534 on Build GRCh38
rs759144415 Chr.19: 12648553-12648553 on Build GRCh38
rs750339878 Chr.19: 12648436-12648437 on Build GRCh38
rs189469608 Chr.19: 12648425-12648425 on Build GRCh38
rs745928175 Chr.19: 12648360-12648360 on Build GRCh38
rs931486929 Chr.19: 12648532-12648532 on Build GRCh38
rs200695002 Chr.19: 12648407-12648407 on Build GRCh38
rs747937275 Chr.19: 12648445-12648445 on Build GRCh38
rs776013861 Chr.19: 12648412-12648412 on Build GRCh38
rs746898354 Chr.19: 12648433-12648433 on Build GRCh38
rs772778684 Chr.19: 12648406-12648406 on Build GRCh38
rs781026700 Chr.19: 12648438-12648438 on Build GRCh38
rs747744816 Chr.19: 12648391-12648391 on Build GRCh38
rs755996633 Chr.19: 12648442-12648443 on Build GRCh38
rs935084853 Chr.19: 12648443-12648443 on Build GRCh38
rs1016249959 Chr.19: 12648522-12648522 on Build GRCh38
rs530923717 Chr.19: 12648460-12648460 on Build GRCh38
rs369174673 Chr.19: 12648374-12648374 on Build GRCh38
rs914908585 Chr.19: 12648458-12648458 on Build GRCh38
rs970921118 Chr.19: 12648386-12648386 on Build GRCh38
rs376734029 Chr.19: 12648428-12648428 on Build GRCh38
rs550855892 Chr.19: 12648424-12648424 on Build GRCh38
rs933863283 Chr.19: 12648489-12648489 on Build GRCh38
rs920051618 Chr.19: 12648514-12648514 on Build GRCh38
rs758456823 Chr.19: 12648426-12648426 on Build GRCh38
rs1021381108 Chr.19: 12648408-12648408 on Build GRCh38
rs764448970 Chr.19: 12648420-12648420 on Build GRCh38
rs561895292 Chr.19: 12648370-12648370 on Build GRCh38
rs1046879893 Chr.19: 12648502-12648502 on Build GRCh38
rs958869615 Chr.19: 12648361-12648361 on Build GRCh38
rs746805353 Chr.19: 12648372-12648372 on Build GRCh38
rs956839427 Chr.19: 12648437-12648437 on Build GRCh38
rs758333025 Chr.19: 12648365-12648365 on Build GRCh38
rs570839557 Chr.19: 12648509-12648509 on Build GRCh38
rs760974907 Chr.19: 12648418-12648418 on Build GRCh38
rs1039625598 Chr.19: 12648470-12648470 on Build GRCh38
rs757044622 Chr.19: 12648429-12648430 on Build GRCh38

View Full Product Details