Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145639041-145639302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
UniGene
Hs.452864
Gene Chromosome Location
Chr.4: 145619388-145660035 on Build GRCh38
Transcripts
NM_172250.2 XM_011531684.2 XM_011531685.2 XM_011531686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
205743 c.129G>A p.P43P Chr.4: 145639268-145639268 on Build GRCh38
377949 c.125A>T p.Q42L Chr.4: 145639264-145639264 on Build GRCh38
6166348 c.52A>G p.R18G Chr.4: 145639191-145639191 on Build GRCh38
4996400 c.89C>T p.S30L Chr.4: 145639228-145639228 on Build GRCh38
367366 c.43G>A p.G15S Chr.4: 145639182-145639182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766651356 Chr.4: 145639137-145639137 on Build GRCh38
rs766309050 Chr.4: 145639215-145639215 on Build GRCh38
rs778693852 Chr.4: 145639176-145639176 on Build GRCh38
rs749162902 Chr.4: 145639091-145639091 on Build GRCh38
rs150065810 Chr.4: 145639255-145639255 on Build GRCh38
rs746718277 Chr.4: 145639192-145639192 on Build GRCh38
rs376276423 Chr.4: 145639095-145639095 on Build GRCh38
rs775224246 Chr.4: 145639189-145639189 on Build GRCh38
rs749076210 Chr.4: 145639171-145639171 on Build GRCh38
rs762276953 Chr.4: 145639216-145639217 on Build GRCh38
rs373547595 Chr.4: 145639111-145639111 on Build GRCh38
rs370709347 Chr.4: 145639207-145639207 on Build GRCh38
rs757224297 Chr.4: 145639173-145639173 on Build GRCh38
rs527340737 Chr.4: 145639146-145639146 on Build GRCh38
rs765799472 Chr.4: 145639203-145639203 on Build GRCh38
rs34502082 Chr.4: 145639073-145639074 on Build GRCh38
rs370983676 Chr.4: 145639172-145639172 on Build GRCh38
rs375682603 Chr.4: 145639204-145639204 on Build GRCh38
rs774967096 Chr.4: 145639103-145639103 on Build GRCh38
rs756348773 Chr.4: 145639218-145639218 on Build GRCh38
rs142126209 Chr.4: 145639231-145639231 on Build GRCh38
rs543412068 Chr.4: 145639276-145639276 on Build GRCh38
rs897695482 Chr.4: 145639267-145639267 on Build GRCh38
rs138947726 Chr.4: 145639126-145639126 on Build GRCh38
rs34702224 Chr.4: 145639277-145639277 on Build GRCh38
rs777599101 Chr.4: 145639159-145639159 on Build GRCh38
rs143211378 Chr.4: 145639196-145639196 on Build GRCh38
rs889048711 Chr.4: 145639206-145639206 on Build GRCh38
rs4835011 Chr.4: 145639084-145639084 on Build GRCh38
rs374109118 Chr.4: 145639136-145639136 on Build GRCh38
rs765634183 Chr.4: 145639131-145639131 on Build GRCh38
rs758937804 Chr.4: 145639102-145639102 on Build GRCh38
rs146372922 Chr.4: 145639241-145639241 on Build GRCh38
rs771417006 Chr.4: 145639184-145639184 on Build GRCh38
rs371810384 Chr.4: 145639183-145639183 on Build GRCh38
rs367809749 Chr.4: 145639225-145639225 on Build GRCh38
rs750568846 Chr.4: 145639135-145639135 on Build GRCh38
rs758345818 Chr.4: 145639263-145639263 on Build GRCh38
rs755914586 Chr.4: 145639154-145639154 on Build GRCh38
rs139395700 Chr.4: 145639251-145639251 on Build GRCh38
rs150259804 Chr.4: 145639105-145639105 on Build GRCh38
rs762251343 Chr.4: 145639197-145639197 on Build GRCh38
rs1027730285 Chr.4: 145639068-145639068 on Build GRCh38
rs751609803 Chr.4: 145639080-145639080 on Build GRCh38
rs746489522 Chr.4: 145639271-145639271 on Build GRCh38
rs921313777 Chr.4: 145639160-145639160 on Build GRCh38
rs745735153 Chr.4: 145639257-145639257 on Build GRCh38
rs893538330 Chr.4: 145639182-145639182 on Build GRCh38
rs886059088 Chr.4: 145639267-145639267 on Build GRCh38
rs765782136 Chr.4: 145639226-145639226 on Build GRCh38
rs779779664 Chr.4: 145639268-145639268 on Build GRCh38

View Full Product Details