Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218275396-218275659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
UniGene
Hs.98475
Gene Chromosome Location
Chr.2: 218270392-218346793 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

Gene Symbol
TMBIM1
Entrez Gene ID
64114
Gene Name
transmembrane BAX inhibitor motif containing 1
Gene Aliases
LFG3, MST100, MSTP100, PP1201, RECS1
UniGene
Hs.591605
Gene Chromosome Location
Chr.2: 218274192-218292577 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
227711 c.923G>C p.G308A Chr.2: 218275488-218275488 on Build GRCh38
304568 c.929G>A p.R310H Chr.2: 218275482-218275482 on Build GRCh38
4598042 c.827G>A p.R276Q Chr.2: 218275584-218275584 on Build GRCh38
5589157 c.802G>T p.D268Y Chr.2: 218275609-218275609 on Build GRCh38
3757952 c.846C>G p.P282P Chr.2: 218275565-218275565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900185550 Chr.2: 218275461-218275461 on Build GRCh38
rs760550492 Chr.2: 218275498-218275499 on Build GRCh38
rs1022380029 Chr.2: 218275586-218275586 on Build GRCh38
rs200810695 Chr.2: 218275534-218275534 on Build GRCh38
rs762204736 Chr.2: 218275463-218275463 on Build GRCh38
rs769454078 Chr.2: 218275542-218275542 on Build GRCh38
rs999545271 Chr.2: 218275618-218275618 on Build GRCh38
rs986958375 Chr.2: 218275431-218275431 on Build GRCh38
rs767298806 Chr.2: 218275447-218275447 on Build GRCh38
rs762590223 Chr.2: 218275454-218275454 on Build GRCh38
rs752481000 Chr.2: 218275485-218275486 on Build GRCh38
rs766550762 Chr.2: 218275483-218275483 on Build GRCh38
rs745698169 Chr.2: 218275538-218275538 on Build GRCh38
rs754073589 Chr.2: 218275576-218275576 on Build GRCh38
rs955509545 Chr.2: 218275417-218275417 on Build GRCh38
rs764689536 Chr.2: 218275457-218275457 on Build GRCh38
rs751447108 Chr.2: 218275585-218275585 on Build GRCh38
rs745668283 Chr.2: 218275609-218275609 on Build GRCh38
rs143173370 Chr.2: 218275482-218275482 on Build GRCh38
rs142976124 Chr.2: 218275547-218275547 on Build GRCh38
rs2292550 Chr.2: 218275565-218275565 on Build GRCh38
rs199711733 Chr.2: 218275478-218275478 on Build GRCh38
rs768237289 Chr.2: 218275637-218275637 on Build GRCh38
rs371106981 Chr.2: 218275632-218275632 on Build GRCh38
rs148573957 Chr.2: 218275561-218275561 on Build GRCh38
rs550448571 Chr.2: 218275479-218275479 on Build GRCh38
rs766610505 Chr.2: 218275571-218275571 on Build GRCh38
rs74675992 Chr.2: 218275436-218275436 on Build GRCh38
rs74826045 Chr.2: 218275559-218275559 on Build GRCh38
rs754982758 Chr.2: 218275583-218275583 on Build GRCh38
rs770725532 Chr.2: 218275447-218275447 on Build GRCh38
rs746952375 Chr.2: 218275516-218275516 on Build GRCh38
rs563258520 Chr.2: 218275433-218275433 on Build GRCh38
rs373791088 Chr.2: 218275595-218275595 on Build GRCh38
rs369057432 Chr.2: 218275452-218275452 on Build GRCh38
rs1031657480 Chr.2: 218275624-218275624 on Build GRCh38
rs755109208 Chr.2: 218275508-218275508 on Build GRCh38
rs376693165 Chr.2: 218275451-218275451 on Build GRCh38
rs148970259 Chr.2: 218275546-218275546 on Build GRCh38
rs377618088 Chr.2: 218275610-218275610 on Build GRCh38
rs781027967 Chr.2: 218275600-218275600 on Build GRCh38
rs748983120 Chr.2: 218275634-218275634 on Build GRCh38
rs903828149 Chr.2: 218275608-218275608 on Build GRCh38
rs777629876 Chr.2: 218275509-218275509 on Build GRCh38
rs149837076 Chr.2: 218275490-218275490 on Build GRCh38
rs11552008 Chr.2: 218275415-218275415 on Build GRCh38
rs762435410 Chr.2: 218275564-218275564 on Build GRCh38
rs781061038 Chr.2: 218275536-218275536 on Build GRCh38
rs752166044 Chr.2: 218275460-218275460 on Build GRCh38

View Full Product Details