Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18786329-18786596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391735 c.1247C>T p.P416L Chr.19: 18786539-18786539 on Build GRCh38
3891978 c.1212G>A p.G404G Chr.19: 18786574-18786574 on Build GRCh38
711091 c.1248G>A p.P416P Chr.19: 18786538-18786538 on Build GRCh38
3422552 c.1213G>A p.D405N Chr.19: 18786573-18786573 on Build GRCh38
438984 c.1191C>T p.D397D Chr.19: 18786595-18786595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764546396 Chr.19: 18786539-18786539 on Build GRCh38
rs368587069 Chr.19: 18786514-18786514 on Build GRCh38
rs764879634 Chr.19: 18786534-18786534 on Build GRCh38
rs147630181 Chr.19: 18786353-18786353 on Build GRCh38
rs909891604 Chr.19: 18786463-18786463 on Build GRCh38
rs368715216 Chr.19: 18786538-18786538 on Build GRCh38
rs746578492 Chr.19: 18786520-18786520 on Build GRCh38
rs770628352 Chr.19: 18786524-18786524 on Build GRCh38
rs886042932 Chr.19: 18786564-18786564 on Build GRCh38
rs375908187 Chr.19: 18786532-18786532 on Build GRCh38
rs1007491344 Chr.19: 18786366-18786366 on Build GRCh38
rs972333523 Chr.19: 18786411-18786411 on Build GRCh38
rs767814536 Chr.19: 18786560-18786560 on Build GRCh38
rs771841538 Chr.19: 18786506-18786506 on Build GRCh38
rs200382113 Chr.19: 18786489-18786489 on Build GRCh38
rs1047499216 Chr.19: 18786516-18786516 on Build GRCh38
rs796435646 Chr.19: 18786423-18786423 on Build GRCh38
rs763339761 Chr.19: 18786363-18786363 on Build GRCh38
rs756200063 Chr.19: 18786569-18786569 on Build GRCh38
rs887239135 Chr.19: 18786523-18786523 on Build GRCh38
rs528720862 Chr.19: 18786358-18786358 on Build GRCh38
rs984081635 Chr.19: 18786456-18786456 on Build GRCh38
rs754834870 Chr.19: 18786496-18786496 on Build GRCh38
rs751860122 Chr.19: 18786540-18786540 on Build GRCh38
rs750654646 Chr.19: 18786562-18786562 on Build GRCh38
rs931273850 Chr.19: 18786437-18786437 on Build GRCh38
rs745984596 Chr.19: 18786490-18786490 on Build GRCh38
rs140712407 Chr.19: 18786370-18786370 on Build GRCh38
rs150080950 Chr.19: 18786401-18786401 on Build GRCh38
rs778795530 Chr.19: 18786499-18786499 on Build GRCh38
rs557014004 Chr.19: 18786553-18786553 on Build GRCh38
rs764590130 Chr.19: 18786537-18786537 on Build GRCh38
rs748137604 Chr.19: 18786501-18786501 on Build GRCh38
rs780165445 Chr.19: 18786570-18786570 on Build GRCh38
rs537701126 Chr.19: 18786419-18786419 on Build GRCh38
rs775953172 Chr.19: 18786527-18786527 on Build GRCh38

View Full Product Details