Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100401838-100402097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
748206 c.1587G>A p.R529R Chr.7: 100401926-100401926 on Build GRCh38
1233252 c.1553A>C p.K518T Chr.7: 100401960-100401960 on Build GRCh38
5698030 c.1545A>C p.L515L Chr.7: 100401968-100401968 on Build GRCh38
3833691 c.1621C>T p.P541S Chr.7: 100401892-100401892 on Build GRCh38
6371215 c.1486G>A p.A496T Chr.7: 100402027-100402027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745640888 Chr.7: 100402066-100402066 on Build GRCh38
rs765960810 Chr.7: 100401896-100401896 on Build GRCh38
rs776911656 Chr.7: 100401967-100401967 on Build GRCh38
rs748813696 Chr.7: 100402062-100402065 on Build GRCh38
rs568467361 Chr.7: 100401914-100401914 on Build GRCh38
rs377758789 Chr.7: 100401897-100401897 on Build GRCh38
rs763250376 Chr.7: 100402010-100402010 on Build GRCh38
rs750883641 Chr.7: 100401881-100401881 on Build GRCh38
rs756108877 Chr.7: 100401927-100401927 on Build GRCh38
rs200816906 Chr.7: 100401969-100401969 on Build GRCh38
rs748253165 Chr.7: 100402078-100402078 on Build GRCh38
rs770706249 Chr.7: 100401957-100401957 on Build GRCh38
rs760711349 Chr.7: 100401924-100401924 on Build GRCh38
rs1033891234 Chr.7: 100401940-100401940 on Build GRCh38
rs754070445 Chr.7: 100402060-100402060 on Build GRCh38
rs755320959 Chr.7: 100402067-100402067 on Build GRCh38
rs766806707 Chr.7: 100402023-100402023 on Build GRCh38
rs375766278 Chr.7: 100401959-100401959 on Build GRCh38
rs536980278 Chr.7: 100402048-100402048 on Build GRCh38
rs752188103 Chr.7: 100401923-100401923 on Build GRCh38
rs754555042 Chr.7: 100401906-100401906 on Build GRCh38
rs374199507 Chr.7: 100402035-100402035 on Build GRCh38
rs778821300 Chr.7: 100402065-100402065 on Build GRCh38
rs764038372 Chr.7: 100402051-100402051 on Build GRCh38
rs569490892 Chr.7: 100402008-100402008 on Build GRCh38
rs775067228 Chr.7: 100402023-100402024 on Build GRCh38
rs763310132 Chr.7: 100401892-100401892 on Build GRCh38
rs914089292 Chr.7: 100402043-100402043 on Build GRCh38
rs200632780 Chr.7: 100402061-100402061 on Build GRCh38
rs187953851 Chr.7: 100402032-100402032 on Build GRCh38
rs780166286 Chr.7: 100401958-100401958 on Build GRCh38
rs755166130 Chr.7: 100402079-100402079 on Build GRCh38
rs372099175 Chr.7: 100402070-100402070 on Build GRCh38
rs1005976675 Chr.7: 100401873-100401873 on Build GRCh38
rs367958253 Chr.7: 100401963-100401963 on Build GRCh38
rs377236763 Chr.7: 100402052-100402052 on Build GRCh38
rs770430680 Chr.7: 100401988-100401988 on Build GRCh38
rs183621422 Chr.7: 100401928-100401928 on Build GRCh38
rs752206215 Chr.7: 100402047-100402047 on Build GRCh38
rs1037510993 Chr.7: 100401875-100401875 on Build GRCh38
rs767150996 Chr.7: 100402038-100402038 on Build GRCh38
rs749047709 Chr.7: 100401930-100401930 on Build GRCh38

View Full Product Details