Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81707169-81707442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6937291 c.1592C>G p.T531S Chr.7: 81707314-81707314 on Build GRCh38
3030139 c.1585G>A p.V529I Chr.7: 81707321-81707321 on Build GRCh38
4168648 c.1558G>A p.G520R Chr.7: 81707348-81707348 on Build GRCh38
6954327 c.1597C>T p.R533* Chr.7: 81707309-81707309 on Build GRCh38
7114218 c.1598G>T p.R533L Chr.7: 81707308-81707308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758387409 Chr.7: 81707254-81707254 on Build GRCh38
rs371139129 Chr.7: 81707281-81707281 on Build GRCh38
rs5745745 Chr.7: 81707219-81707219 on Build GRCh38
rs747804909 Chr.7: 81707271-81707271 on Build GRCh38
rs764035851 Chr.7: 81707343-81707343 on Build GRCh38
rs751552963 Chr.7: 81707405-81707405 on Build GRCh38
rs541629166 Chr.7: 81707395-81707395 on Build GRCh38
rs895204415 Chr.7: 81707212-81707212 on Build GRCh38
rs368519009 Chr.7: 81707292-81707292 on Build GRCh38
rs1011014142 Chr.7: 81707218-81707218 on Build GRCh38
rs779909459 Chr.7: 81707255-81707255 on Build GRCh38
rs771437724 Chr.7: 81707308-81707308 on Build GRCh38
rs750290140 Chr.7: 81707249-81707249 on Build GRCh38
rs535867555 Chr.7: 81707207-81707207 on Build GRCh38
rs751391507 Chr.7: 81707261-81707261 on Build GRCh38
rs761860902 Chr.7: 81707388-81707388 on Build GRCh38
rs764242287 Chr.7: 81707241-81707241 on Build GRCh38
rs941486309 Chr.7: 81707214-81707214 on Build GRCh38
rs370927722 Chr.7: 81707267-81707267 on Build GRCh38
rs527951814 Chr.7: 81707309-81707309 on Build GRCh38
rs776754331 Chr.7: 81707353-81707353 on Build GRCh38
rs745956310 Chr.7: 81707256-81707256 on Build GRCh38
rs150272458 Chr.7: 81707268-81707268 on Build GRCh38
rs143262934 Chr.7: 81707328-81707328 on Build GRCh38
rs759881207 Chr.7: 81707312-81707312 on Build GRCh38
rs888713835 Chr.7: 81707213-81707213 on Build GRCh38
rs376013029 Chr.7: 81707415-81707415 on Build GRCh38
rs775625219 Chr.7: 81707325-81707325 on Build GRCh38
rs748796647 Chr.7: 81707307-81707307 on Build GRCh38
rs772441910 Chr.7: 81707316-81707316 on Build GRCh38
rs905243504 Chr.7: 81707246-81707246 on Build GRCh38

View Full Product Details