Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52448771-52449044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
UniGene
Hs.708950
Gene Chromosome Location
Chr.12: 52446651-52452126 on Build GRCh38
Transcripts
NM_005555.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135395 c.1200G>T p.K400N Chr.12: 52448845-52448845 on Build GRCh38
3736061 c.1186G>A p.D396N Chr.12: 52448859-52448859 on Build GRCh38
1362491 c.1191C>T p.H397H Chr.12: 52448854-52448854 on Build GRCh38
940723 c.1123C>T p.R375C Chr.12: 52448922-52448922 on Build GRCh38
3936017 c.1167G>T p.Q389H Chr.12: 52448878-52448878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11170125 Chr.12: 52448978-52448978 on Build GRCh38
rs755221849 Chr.12: 52448813-52448813 on Build GRCh38
rs202219327 Chr.12: 52449003-52449003 on Build GRCh38
rs750950932 Chr.12: 52448998-52448998 on Build GRCh38
rs771241058 Chr.12: 52448838-52448838 on Build GRCh38
rs774203775 Chr.12: 52448882-52448882 on Build GRCh38
rs767317812 Chr.12: 52448809-52448809 on Build GRCh38
rs757806566 Chr.12: 52448924-52448924 on Build GRCh38
rs368362440 Chr.12: 52448888-52448888 on Build GRCh38
rs765512406 Chr.12: 52448901-52448901 on Build GRCh38
rs756540663 Chr.12: 52448928-52448928 on Build GRCh38
rs200590482 Chr.12: 52448807-52448807 on Build GRCh38
rs437014 Chr.12: 52448952-52448952 on Build GRCh38
rs768268060 Chr.12: 52448860-52448860 on Build GRCh38
rs372419655 Chr.12: 52448989-52448989 on Build GRCh38
rs773938726 Chr.12: 52448945-52448945 on Build GRCh38
rs762849989 Chr.12: 52448807-52448807 on Build GRCh38
rs766144450 Chr.12: 52448972-52448972 on Build GRCh38
rs768087354 Chr.12: 52448942-52448942 on Build GRCh38
rs779910657 Chr.12: 52448929-52448929 on Build GRCh38
rs112322706 Chr.12: 52448840-52448840 on Build GRCh38
rs568733370 Chr.12: 52448800-52448800 on Build GRCh38
rs746172336 Chr.12: 52448851-52448851 on Build GRCh38
rs772057116 Chr.12: 52448960-52448960 on Build GRCh38
rs146904435 Chr.12: 52448854-52448854 on Build GRCh38
rs374325319 Chr.12: 52448994-52448994 on Build GRCh38
rs764173617 Chr.12: 52448984-52448984 on Build GRCh38
rs747532177 Chr.12: 52448835-52448835 on Build GRCh38
rs778205606 Chr.12: 52448833-52448833 on Build GRCh38
rs758471569 Chr.12: 52448831-52448831 on Build GRCh38
rs767313923 Chr.12: 52448889-52448889 on Build GRCh38
rs751510770 Chr.12: 52448804-52448804 on Build GRCh38
rs760524758 Chr.12: 52448966-52448966 on Build GRCh38
rs748925772 Chr.12: 52448936-52448936 on Build GRCh38
rs773022037 Chr.12: 52448965-52448965 on Build GRCh38
rs139792514 Chr.12: 52448847-52448847 on Build GRCh38
rs138988810 Chr.12: 52448859-52448859 on Build GRCh38
rs760398334 Chr.12: 52448890-52448890 on Build GRCh38
rs751554113 Chr.12: 52448922-52448922 on Build GRCh38
rs779055245 Chr.12: 52448823-52448823 on Build GRCh38
rs113429102 Chr.12: 52448841-52448841 on Build GRCh38
rs1026981742 Chr.12: 52448829-52448829 on Build GRCh38
rs746604930 Chr.12: 52448993-52448994 on Build GRCh38
rs376000522 Chr.12: 52448996-52448996 on Build GRCh38
rs775079590 Chr.12: 52448983-52448984 on Build GRCh38
rs772147724 Chr.12: 52448799-52448800 on Build GRCh38
rs200904701 Chr.12: 52449004-52449004 on Build GRCh38
rs144784042 Chr.12: 52448800-52448800 on Build GRCh38
rs756595848 Chr.12: 52448999-52448999 on Build GRCh38
rs775733295 Chr.12: 52448973-52448973 on Build GRCh38
rs781494966 Chr.12: 52448927-52448927 on Build GRCh38
rs752907331 Chr.12: 52448911-52448911 on Build GRCh38
rs374251478 Chr.12: 52448815-52448815 on Build GRCh38
rs960455848 Chr.12: 52448862-52448862 on Build GRCh38
rs762492971 Chr.12: 52448795-52448795 on Build GRCh38
rs764203649 Chr.12: 52448921-52448921 on Build GRCh38
rs758526690 Chr.12: 52448913-52448913 on Build GRCh38
rs752780925 Chr.12: 52448828-52448828 on Build GRCh38
rs370922722 Chr.12: 52448812-52448812 on Build GRCh38

View Full Product Details