Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23919187-23919442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL15
Entrez Gene ID
6138
Gene Name
ribosomal protein L15
Gene Aliases
DBA12, EC45, L15, RPL10, RPLY10, RPYL10
UniGene
Hs.381219
Gene Chromosome Location
Chr.3: 23916804-23923696 on Build GRCh38
Transcripts
NM_001253379.1 NM_001253380.1 NM_001253382.1 NM_001253383.1 NM_001253384.1 NM_002948.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329938 c.342A>G p.R114R Chr.3: 23919228-23919228 on Build GRCh38
6329939 c.397A>T p.I133F Chr.3: 23919283-23919283 on Build GRCh38
5183719 c.324C>T p.R108R Chr.3: 23919210-23919210 on Build GRCh38
6329937 c.317C>T p.A106V Chr.3: 23919203-23919203 on Build GRCh38
5183720 c.468C>T p.H156H Chr.3: 23919354-23919354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151218632 Chr.3: 23919267-23919267 on Build GRCh38
rs768075641 Chr.3: 23919327-23919327 on Build GRCh38
rs11547051 Chr.3: 23919270-23919270 on Build GRCh38
rs377507939 Chr.3: 23919310-23919310 on Build GRCh38
rs748865914 Chr.3: 23919417-23919417 on Build GRCh38
rs886242247 Chr.3: 23919300-23919300 on Build GRCh38
rs534301057 Chr.3: 23919325-23919325 on Build GRCh38
rs181790084 Chr.3: 23919381-23919381 on Build GRCh38
rs373573769 Chr.3: 23919363-23919363 on Build GRCh38
rs554703410 Chr.3: 23919419-23919419 on Build GRCh38
rs757540488 Chr.3: 23919354-23919354 on Build GRCh38
rs753203979 Chr.3: 23919328-23919328 on Build GRCh38
rs777381939 Chr.3: 23919401-23919401 on Build GRCh38
rs370700905 Chr.3: 23919344-23919344 on Build GRCh38
rs768764156 Chr.3: 23919227-23919227 on Build GRCh38
rs769440685 Chr.3: 23919262-23919262 on Build GRCh38
rs766621970 Chr.3: 23919288-23919288 on Build GRCh38
rs747729430 Chr.3: 23919249-23919249 on Build GRCh38
rs748116544 Chr.3: 23919391-23919391 on Build GRCh38
rs3088096 Chr.3: 23919226-23919226 on Build GRCh38
rs769387719 Chr.3: 23919392-23919392 on Build GRCh38
rs558281601 Chr.3: 23919225-23919225 on Build GRCh38
rs1023749007 Chr.3: 23919399-23919399 on Build GRCh38
rs759943814 Chr.3: 23919326-23919326 on Build GRCh38
rs758083776 Chr.3: 23919217-23919217 on Build GRCh38
rs776268885 Chr.3: 23919231-23919231 on Build GRCh38
rs756131448 Chr.3: 23919333-23919333 on Build GRCh38
rs781215865 Chr.3: 23919387-23919387 on Build GRCh38
rs3211002 Chr.3: 23919383-23919383 on Build GRCh38
rs998038229 Chr.3: 23919210-23919210 on Build GRCh38
rs753971737 Chr.3: 23919346-23919346 on Build GRCh38
rs774230761 Chr.3: 23919269-23919269 on Build GRCh38
rs779933912 Chr.3: 23919218-23919218 on Build GRCh38
rs749541129 Chr.3: 23919209-23919209 on Build GRCh38
rs746880398 Chr.3: 23919375-23919375 on Build GRCh38
rs373453626 Chr.3: 23919309-23919309 on Build GRCh38

View Full Product Details