Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1351684-1351909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVSSA
Entrez Gene ID
57654
Gene Name
UV stimulated scaffold protein A
Gene Aliases
KIAA1530, UVSS3
UniGene
Hs.380475
Gene Chromosome Location
Chr.4: 1345662-1388049 on Build GRCh38
Transcripts
NM_001317934.1 NM_001317935.1 NM_020894.3 XM_017008490.1 XM_017008491.1 XM_017008492.1 XM_017008493.1 XM_017008494.1 XM_017008495.1 XM_017008496.1 XM_017008497.1 XM_017008498.1 XM_017008499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3825200 c.509T>G p.I170S Chr.4: 1351794-1351794 on Build GRCh38
3428166 c.447G>A p.T149T Chr.4: 1351732-1351732 on Build GRCh38
4740056 c.507A>C p.K169N Chr.4: 1351792-1351792 on Build GRCh38
3010564 c.517G>C p.E173Q Chr.4: 1351802-1351802 on Build GRCh38
4740057 c.523G>A p.A175T Chr.4: 1351808-1351808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184543158 Chr.4: 1351883-1351883 on Build GRCh38
rs752314455 Chr.4: 1351869-1351869 on Build GRCh38
rs779952266 Chr.4: 1351726-1351726 on Build GRCh38
rs201414673 Chr.4: 1351749-1351749 on Build GRCh38
rs775656470 Chr.4: 1351832-1351832 on Build GRCh38
rs746356055 Chr.4: 1351823-1351823 on Build GRCh38
rs766354705 Chr.4: 1351862-1351862 on Build GRCh38
rs201407968 Chr.4: 1351819-1351819 on Build GRCh38
rs767741018 Chr.4: 1351753-1351753 on Build GRCh38
rs138228745 Chr.4: 1351729-1351729 on Build GRCh38
rs200745723 Chr.4: 1351769-1351769 on Build GRCh38
rs10000797 Chr.4: 1351859-1351859 on Build GRCh38
rs373828028 Chr.4: 1351818-1351818 on Build GRCh38
rs200163688 Chr.4: 1351789-1351789 on Build GRCh38
rs553111337 Chr.4: 1351713-1351713 on Build GRCh38
rs760963274 Chr.4: 1351764-1351764 on Build GRCh38
rs111997159 Chr.4: 1351866-1351866 on Build GRCh38
rs751291436 Chr.4: 1351865-1351865 on Build GRCh38
rs765203388 Chr.4: 1351858-1351858 on Build GRCh38
rs200811036 Chr.4: 1351781-1351781 on Build GRCh38
rs879744855 Chr.4: 1351788-1351788 on Build GRCh38
rs775989117 Chr.4: 1351758-1351758 on Build GRCh38
rs180864391 Chr.4: 1351877-1351877 on Build GRCh38
rs141095512 Chr.4: 1351739-1351739 on Build GRCh38
rs747671544 Chr.4: 1351740-1351740 on Build GRCh38
rs542457962 Chr.4: 1351796-1351796 on Build GRCh38
rs768367735 Chr.4: 1351731-1351731 on Build GRCh38
rs1003606043 Chr.4: 1351737-1351737 on Build GRCh38
rs771361310 Chr.4: 1351748-1351748 on Build GRCh38
rs772692402 Chr.4: 1351826-1351826 on Build GRCh38
rs772816983 Chr.4: 1351822-1351822 on Build GRCh38
rs750308478 Chr.4: 1351785-1351785 on Build GRCh38
rs367981319 Chr.4: 1351844-1351844 on Build GRCh38
rs1045079695 Chr.4: 1351728-1351728 on Build GRCh38
rs758153489 Chr.4: 1351716-1351716 on Build GRCh38
rs762178075 Chr.4: 1351857-1351857 on Build GRCh38
rs143198789 Chr.4: 1351809-1351809 on Build GRCh38
rs956956849 Chr.4: 1351734-1351734 on Build GRCh38
rs781124970 Chr.4: 1351766-1351766 on Build GRCh38
rs781012577 Chr.4: 1351868-1351868 on Build GRCh38
rs765220920 Chr.4: 1351783-1351783 on Build GRCh38
rs533430571 Chr.4: 1351888-1351888 on Build GRCh38
rs372089831 Chr.4: 1351850-1351850 on Build GRCh38
rs764421923 Chr.4: 1351767-1351767 on Build GRCh38
rs777011965 Chr.4: 1351854-1351854 on Build GRCh38
rs376973809 Chr.4: 1351851-1351851 on Build GRCh38
rs368797236 Chr.4: 1351751-1351751 on Build GRCh38
rs986154883 Chr.4: 1351759-1351759 on Build GRCh38
rs761121326 Chr.4: 1351846-1351846 on Build GRCh38
rs758257857 Chr.4: 1351861-1351861 on Build GRCh38
rs779713360 Chr.4: 1351793-1351793 on Build GRCh38

View Full Product Details