The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004259.6 | NP_004250.4 | 11 - 12 | 1750 | 74 |
XM_005257818.3 | XP_005257875.1 | 11 - 12 | 1763 | 74 | |
XM_005257822.3 | XP_005257879.1 | 4 - 5 | 1121 | 74 | |
XM_005257823.3 | XP_005257880.1 | 11 - 12 | 1762 | 74 | |
XM_006722186.2 | XP_006722249.1 | 4 - 5 | 448 | 74 | |
XM_011525482.2 | XP_011523784.1 | 10 - 11 | 2230 | 74 | |
XM_011525484.1 | XP_011523786.1 | 5 - 6 | 705 | 74 | |
XM_011525485.2 | XP_011523787.1 | 5 - 6 | 644 | 74 | |
XM_011525486.2 | XP_011523788.1 | 4 - 5 | 631 | 74 | |
XM_017025343.1 | XP_016880832.1 | 10 - 11 | 2230 | 74 | |
GenBank mRNA | AB042824.1 | - | 11 - 12 | 1750 | 74 |
AF193041.1 | - | 4 - 5 | 1126 | 74 | |
AK304168.1 | - | 11 - 12 | 1742 | 74 | |
BC000570.2 | - | 5 - 6 | 515 | 74 | |
BC063440.1 | - | 11 - 12 | 1622 | 74 | |
EF560729.1 | - | 11 - 12 | 1715 | 74 |