The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006097.4 | NP_006088.2 | 2 - 3 | 288 | 64 |
GenBank mRNA | AF176042.1 | - | 1 - 2 | 224 | 64 |
AK130879.1 | - | 2 - 3 | 253 | 64 | |
AL697699.1 | - | 2 - 3 | 64 | ||
BQ953325.1 | - | - | 278 | 64 | |
CR456843.1 | - | 1 - 2 | 184 | 64 | |
D82057.1 | - | 2 - 3 | 256 | 64 | |
J02854.1 | - | 1 - 2 | 226 | 64 |