Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF8
Entrez Gene ID
23133
Gene Name
PHD finger protein 8
Gene Aliases
JHDM1F, MRXSSD, ZNF422
Chromosome Location
Chr.X: 53936680 - 54048935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184896.1 NP_001171825.1 8 - 9 1301 97
NM_001184897.1 NP_001171826.1 8 - 9 1452 97
NM_001184898.1 NP_001171827.1 8 - 9 1123 97
NM_015107.2 NP_055922.1 8 - 9 1452 97
XM_005261996.1 XP_005262053.1 9 - 10 1145 97
XM_005261997.3 XP_005262054.1 8 - 9 1750 97
XM_005261999.1 XP_005262056.1 8 - 9 1121 97
XM_005262000.1 XP_005262057.1 8 - 9 1414 97
XM_011530778.1 XP_011529080.1 8 - 9 1414 97
XM_017029361.1 XP_016884850.1 8 - 9 1700 97
XM_017029362.1 XP_016884851.1 8 - 9 1700 97
GenBank mRNA AB029034.1 - 8 - 9 1128 97
AF091081.1 - 2 - 3 245 97
AK021696.1 - 7 - 8 956 97
AK304272.1 - 8 - 9 1274 97
BC053861.1 - 8 - 9 1156 97

View Full Product Details