Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKAG2
Entrez Gene ID
51422
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location
Chr.7: 151556114 - 151877231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040633.1 NP_001035723.1 10 - 11 1447 123
NM_001304527.1 NP_001291456.1 8 - 9 1051 123
NM_001304531.1 NP_001291460.1 7 - 8 469 123
NM_016203.3 NP_057287.2 10 - 11 1716 123
NM_024429.1 NP_077747.1 6 - 7 541 123
XM_005250002.3 XP_005250059.1 10 - 11 1582 123
XM_005250004.3 XP_005250061.1 10 - 11 1528 123
XM_005250006.4 XP_005250063.1 8 - 9 1281 123
XM_006716021.2 XP_006716084.1 10 - 11 1119 123
XM_011516282.1 XP_011514584.1 10 - 11 1116 123
XM_011516283.1 XP_011514585.1 10 - 11 1119 123
XM_011516284.1 XP_011514586.1 10 - 11 1116 123
XM_011516285.1 XP_011514587.1 7 - 8 961 123
XM_011516286.1 XP_011514588.1 6 - 7 482 123
XM_011516287.1 XP_011514589.1 9 - 10 1192 123
XM_017012268.1 XP_016867757.1 10 - 11 1525 123
XM_017012269.1 XP_016867758.1 10 - 11 1330 123
XM_017012270.1 XP_016867759.1 10 - 11 1106 123
XM_017012271.1 XP_016867760.1 10 - 11 1525 123
XM_017012272.1 XP_016867761.1 10 - 11 1103 123
XM_017012273.1 XP_016867762.1 8 - 9 1266 123
XM_017012274.1 XP_016867763.1 7 - 8 958 123
XM_017012275.1 XP_016867764.1 9 - 10 1455 123
XM_017012276.1 XP_016867765.1 10 - 11 1551 123
XM_017012277.1 XP_016867766.1 6 - 7 482 123
XM_017012278.1 XP_016867767.1 9 - 10 1192 123
XM_017012279.1 XP_016867768.1 9 - 10 1458 123
XM_017012280.1 XP_016867769.1 9 - 10 1455 123
XM_017012281.1 XP_016867770.1 9 - 10 1481 123
XM_017012282.1 XP_016867771.1 8 - 9 901 123
GenBank mRNA AB025580.1 - 6 - 7 454 123
AF087875.1 - 6 - 7 502 123
AJ249976.1 - 10 - 11 1195 123
AK001887.1 - 6 - 7 541 123
AK301123.1 - 8 - 9 123
AK307798.1 - 9 - 10 1263 123
AK308116.1 - 10 - 11 1147 123
AK314646.1 - 6 - 7 503 123
AW664063.1 - 7 - 8 441 123
BC020540.1 - 6 - 7 422 123
BC068598.1 - 10 - 11 1416 123
BT007127.1 - 6 - 7 382 123
DA151645.1 - 7 - 8 470 123

View Full Product Details