The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020799.3 | NP_065850.1 | 4 - 5 | 832 | 86 |
XM_006717928.2 | XP_006717991.1 | 5 - 6 | 933 | 86 | |
XM_006717930.3 | XP_006717993.1 | 4 - 5 | 1420 | 86 | |
XM_011539983.1 | XP_011538285.1 | 5 - 6 | 1340 | 86 | |
XM_011539985.1 | XP_011538287.1 | 5 - 6 | 663 | 86 | |
XM_017016453.1 | XP_016871942.1 | 5 - 6 | 1657 | 86 | |
XM_017016454.1 | XP_016871943.1 | 6 - 7 | 1141 | 86 | |
XM_017016455.1 | XP_016871944.1 | 7 - 8 | 1596 | 86 | |
XM_017016456.1 | XP_016871945.1 | 5 - 6 | 1253 | 86 | |
XM_017016457.1 | XP_016871946.1 | 4 - 5 | 1179 | 86 | |
XM_017016458.1 | XP_016871947.1 | 2 - 3 | 357 | 86 | |
GenBank mRNA | AB010120.1 | - | 3 - 4 | 360 | 86 |
AB037794.1 | - | 4 - 5 | 1153 | 86 | |
AK056086.1 | - | 4 - 5 | 761 | 86 | |
AK092946.1 | - | 3 - 4 | 961 | 86 | |
BC010846.1 | - | 4 - 5 | 750 | 86 | |
BX649096.1 | - | 5 - 6 | 9255 | 86 |