The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289088.1 | NP_001276017.1 | 10 - 11 | 1581 | 114 |
NM_001289089.1 | NP_001276018.1 | 9 - 10 | 1240 | 114 | |
NM_001289090.1 | NP_001276019.1 | 10 - 11 | 1345 | 114 | |
NM_001289091.1 | NP_001276020.1 | 10 - 11 | 1415 | 114 | |
NM_001319955.1 | NP_001306884.1 | 8 - 9 | 1127 | 114 | |
NM_024772.4 | NP_079048.3 | 9 - 10 | 1297 | 114 | |
XM_011542163.2 | XP_011540465.1 | 8 - 9 | 1179 | 114 | |
XM_011542165.2 | XP_011540467.1 | 10 - 11 | 1360 | 114 | |
XM_011542166.2 | XP_011540468.1 | 9 - 10 | 1263 | 114 | |
XM_017002358.1 | XP_016857847.1 | 7 - 8 | 1071 | 114 | |
XM_017002359.1 | XP_016857848.1 | 8 - 9 | 1088 | 114 | |
XM_017002360.1 | XP_016857849.1 | 10 - 11 | 1354 | 114 | |
XM_017002361.1 | XP_016857850.1 | 3 - 4 | 210 | 114 | |
GenBank mRNA | AK096206.1 | - | 10 - 11 | 1393 | 114 |
AK125741.1 | - | 10 - 11 | 1581 | 114 | |
AK299779.1 | - | 9 - 10 | 1240 | 114 | |
BX537537.1 | - | 9 - 10 | 1269 | 114 | |
BX648307.1 | - | 8 - 9 | 1100 | 114 |